Skip to Content
Merck
All Photos(1)

Key Documents

W429300

Sigma-Aldrich

1-Octene

98%

Synonym(s):

1-caprylene, Hexylethylene, Oct-1-ene, 1-octylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH=CH2
CAS Number:
Molecular Weight:
112.21
FEMA Number:
4293
Beilstein:
1734497
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.070
NACRES:
NA.21
Organoleptic:
camphoraceous
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

vapor density

3.9 (vs air)

vapor pressure

36 mmHg ( 38 °C)

Assay

98%

form

liquid

autoignition temp.

446 °F

expl. lim.

3.9 %

refractive index

n20/D 1.408 (lit.)
n20/D 1.409 (lit.)

bp

122-123 °C (lit.)

mp

−101 °C (lit.)

density

0.715 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

camphoraceous

SMILES string

CCCCCCC=C

InChI

1S/C8H16/c1-3-5-7-8-6-4-2/h3H,1,4-8H2,2H3

InChI key

KWKAKUADMBZCLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Octene is an unsaturated compound that is not directly used as a food additive but may be used as an indirect food additive in the production of food contact materials such as packaging, containers, and other materials that encounter food during processing, packaging, transportation, and storage .

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Linear low density polyethylene: Microstructure and sealing properties correlation
Simanke A, et al.
Polymer Testing, 32(2), 279-290 (2013)
Sara M Aschmann et al.
Environmental science & technology, 44(10), 3825-3831 (2010-04-28)
Products of the gas-phase reactions of OH radicals with 1-octene and 7-tetradecene have been investigated at 296 +/- 2 K and atmospheric pressure of air, using gas chromatography, direct air sampling atmospheric pressure ionization tandem mass spectrometry, and in situ
A Hempel-Jørgensen et al.
Archives of environmental health, 54(2), 86-94 (1999-03-27)
In this study, we investigated the time course effect of sensory eye irritation in 16 subjects exposed (i.e., eye only) to n-butanol and 1-octene. Half the subjects were exposed to n-butanol, and the remaining subjects were exposed to 1-octene. Each
F P Guengerich
Biochemical and biophysical research communications, 138(1), 193-198 (1986-07-16)
The heme in rat liver microsomal cytochrome P-450 was labeled with 14C or 3H and the microsomes were fractionated after in vitro incubations with a variety of agents known to destroy cytochrome P-450 heme. A major fraction of the heme
Mark Kuil et al.
Journal of the American Chemical Society, 128(35), 11344-11345 (2006-08-31)
We report the formation of high-precision catalysts using encapsulated rhodium complexes. In the current example, the encapsulated rhodium catalyst shows unprecedented high selectivity in the rhodium-catalyzed hydroformylation of internal alkenes, forming predominantly one of the branched aldehydes. This catalyst system

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service