Skip to Content
Merck
All Photos(1)

Key Documents

W373702

Sigma-Aldrich

Vanillyl alcohol

≥98%, FG

Synonym(s):

4-Hydroxy-3-methoxybenzyl alcohol, NSC 3993, Vanillyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH2OH
CAS Number:
Molecular Weight:
154.16
FEMA Number:
3737
Beilstein:
1910044
EC Number:
Council of Europe no.:
690
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.213
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

description

Natural occurrence: Beer

Assay

≥98%

mp

110-117 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy; sweet; vanilla

SMILES string

COc1cc(CO)ccc1O

InChI

1S/C8H10O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-4,9-10H,5H2,1H3

InChI key

ZENOXNGFMSCLLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Possible uses: vanilla, coconut, cream and other dairy nuances, coumarin notes.

Biochem/physiol Actions

Odor at 1.0%: sweet creamy, phenolic, vanilla and coconut-like with slight brown and coumarinic nuances.
Taste at 50 ppm: creamy waxy, sweet milky, powdery with an ice cream-like nuance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, Gerard
Perfumer & Flavorist, 34, 51-51 (2009)
Kiki L Larsen et al.
The journal of physical chemistry. B, 115(39), 11470-11480 (2011-08-13)
Structural analysis of plant materials, i.e., lignin, cellulose, hemicellulose, etc., supports the growing interest of their uses, e.g., as sources for biofuels or materials. Lignin is a main polymer formed from three phenolic presursors, containing none, one, or two OMe
In Su Kim et al.
Molecules (Basel, Switzerland), 16(7), 5349-5361 (2011-06-28)
Gastrodia elata Blume (GE) has long been used in oriental countries as a traditional herbal medicine to relieve symptoms associated with neurological ailments such as vertigo, general paralysis and epilepsy. In this study, we have investigated the effects of GE
Padmesh Venkitasubramanian et al.
The Journal of biological chemistry, 282(1), 478-485 (2006-11-15)
Aldehyde oxidoreductase (carboxylic acid reductase (Car)) catalyzes the magnesium-, ATP-, and NADPH-dependent reduction of carboxylic acids to their corresponding aldehydes. Heterologous expression of the car gene in Escherichia coli afforded purified recombinant enzyme with a specific activity nearly 50-fold lower
C L Hsieh et al.
Life sciences, 67(10), 1185-1195 (2000-08-23)
Vanillyl alcohol (VA) is a component of Gastrodia elata Bl. (GE), which is a traditional Chinese herb widely used to treat convulsive disorders or dizziness. This study examined the role of VA in the anticonvulsive properties of GE in a

Articles

Vanilla Regulations – The Law and the Labeling of the World’s Most Popular Flavor

Vanilla Regulations – The Law and the Labeling of the World’s Most Popular Flavor

Vanilla Regulations – The Law and the Labeling of the World’s Most Popular Flavor

Vanilla Regulations – The Law and the Labeling of the World’s Most Popular Flavor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service