Skip to Content
Merck
All Photos(1)

Key Documents

W344400

Sigma-Aldrich

DL-Valine

≥97%

Synonym(s):

(±)-α-Aminoisovaleric acid, DL-2-Amino-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(NH2)COOH
CAS Number:
Molecular Weight:
117.15
FEMA Number:
3444
Beilstein:
506689
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
17.023
NACRES:
NA.21
Organoleptic:
odorless
grade:
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Halal

Assay

≥97%

mp

295 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

CC(C)C(N)C(O)=O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)

InChI key

KZSNJWFQEVHDMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksander Hinek et al.
Journal of dermatological science, 75(3), 173-182 (2014-07-13)
Vitamin C (L-ascorbic acid), a known enhancer of collagen deposition, has also been identified as an inhibitor of elastogenesis. Present studies explored whether and how the L-ascorbic acid derivative (+) sodium L-ascorbate (SA) would affect production of collagen and elastic
Yan-Mei Ma et al.
Fish & shellfish immunology, 45(2), 414-421 (2015-05-11)
Streptococcus iniae seriously affects the intensive farming of tilapias. Much work has been conducted on prevention and control of S. iniae infection, but little published information on the metabolic response is available in tilapias against the bacterial infection, and no metabolic
K Aaboe et al.
Diabetes, obesity & metabolism, 17(1), 74-81 (2014-09-23)
To examine whether 12 weeks of treatment with a dipeptidyl peptidase-4 (DPP-4) inhibitor, sitagliptin, influences the insulin secretion induced by glucose, glucose-dependent insulinotropic polypeptide (GIP) and glucagon-like peptide-1 (GLP-1) during a hyperglycaemic clamp in patients with type 2 diabetes (T2DM).
Thomas Rydzak et al.
Journal of industrial microbiology & biotechnology, 42(9), 1263-1272 (2015-07-15)
The ability of Clostridium thermocellum to rapidly degrade cellulose and ferment resulting hydrolysis products into ethanol makes it a promising platform organism for cellulosic biofuel production via consolidated bioprocessing. Currently, however, ethanol yield is far below theoretical maximum due to
Yuka Kamei et al.
The Journal of biological chemistry, 289(46), 32081-32093 (2014-10-09)
Age-related damage accumulates and a variety of biological activities and functions deteriorate in senescent cells. However, little is known about when cellular aging behaviors begin and what cellular aging processes change. Previous research demonstrated age-related mRNA changes in budding yeast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service