Skip to Content
Merck
All Photos(1)

Key Documents

W324108

Sigma-Aldrich

Trimethylamine solution

25 wt. % in H2O

Synonym(s):

N,N-dimethylmethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N
CAS Number:
Molecular Weight:
59.11
FEMA Number:
3241
Beilstein:
956566
EC Number:
Council of Europe no.:
10497
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
11.009
NACRES:
NA.21
Organoleptic:
fishy; meaty; oily; fruity; rancid
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Kosher

vapor density

2.04 (vs air)

vapor pressure

232 mmHg ( 20 °C)

autoignition temp.

374 °F

concentration

25 wt. % in H2O

refractive index

n20/D 1.357

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fishy; meaty; oily; fruity; rancid

SMILES string

CN(C)C

InChI

1S/C3H9N/c1-4(2)3/h1-3H3

InChI key

GETQZCLCWQTVFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Unraveling Ammonia and Trimethylamine Uptake on Conductive Doped Polyaniline.: This research investigates the adsorption properties of ammonia and trimethylamine on conductive doped polyaniline, contributing to the development of advanced materials for gas sensing applications (Pascaud et al., 2024).

  • TMAO perturbs intermolecular vibrational motions of water revealed by low-frequency modes.: The study delves into how trimethylamine-N-oxide (TMAO) affects the vibrational motions of water molecules, providing insights into the molecular interactions in aqueous solutions (Liu & Okuno, 2024).

  • Trimethylamine-N-oxide depletes urea in a peptide solvation shell.: This article explores the interaction between TMAO and urea in peptide solvation shells, enhancing our understanding of protein stability in varying chemical environments (Nasralla et al., 2024).

  • Co-Solutes Induced Changes in the Properties of Polymeric Solution and Water Dynamics.: This research investigates the impact of co-solutes, including trimethylamine, on the properties of polymeric solutions and water dynamics, contributing to the field of solution chemistry (Bhattacharjee et al., 2024).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

37.9 °F - closed cup

Flash Point(C)

3.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abhishek Bhattacharya et al.
Cell, 176(5), 1174-1189 (2019-01-29)
The specific patterns and functional properties of electrical synapses of a nervous system are defined by the neuron-specific complement of electrical synapse constituents. We systematically examined the molecular composition of the electrical connectome of the nematode C. elegans through a genome-
S L Hillier
American journal of obstetrics and gynecology, 169(2 Pt 2), 455-459 (1993-08-01)
Bacterial vaginosis is characterized microbiologically by replacement of the Lactobacillus-predominant vaginal flora by Gardnerella vaginalis, Bacteroides species, Mobiluncus species, and genital mycoplasmas. A standardized, laboratory-based diagnostic test for bacterial vaginosis is desirable in those instances in which a microscope is
M Al-Waiz et al.
Clinical pharmacology and therapeutics, 42(5), 588-594 (1987-11-01)
Trimethylamine (TMA) and its N-oxide (TMAO) are normal components of human urine. They are present in the diet and also derived from the enterobacterial metabolism of precursors such as choline. Dietary TMA is almost entirely metabolized to and excreted as
Hyeryeong Jeon et al.
Macromolecular rapid communications, 33(11), 972-976 (2012-04-12)
Polydiacetylenes (PDAs), a family of conjugated polymers, are very intriguing materials in several aspects. Especially, the stimulus-induced apparent blue-to-red transition of the PDAs has led to the development of a variety of PDA-based chemosensors. In the current work, we synthesized
Qian Li et al.
Current biology : CB, 23(1), 11-20 (2012-11-28)
Rodents use olfactory cues for species-specific behaviors. For example, mice emit odors to attract mates of the same species, but not competitors of closely related species. This implies rapid evolution of olfactory signaling, although odors and chemosensory receptors involved are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service