Skip to Content
Merck
All Photos(1)

Key Documents

W267511

Sigma-Aldrich

2-Methoxy-4-vinylphenol

≥98%, FG

Synonym(s):

4-Vinyl guaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CH=CH2)OH
CAS Number:
Molecular Weight:
150.17
FEMA Number:
2675
EC Number:
Council of Europe no.:
177
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.009
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.582 (lit.)

bp

224 °C (lit.)

density

1.11 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cedar; woody; peanut

SMILES string

COc1cc(C=C)ccc1O

InChI

1S/C9H10O2/c1-3-7-4-5-8(10)9(6-7)11-2/h3-6,10H,1H2,2H3

InChI key

YOMSJEATGXXYPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Methoxy-4-vinylphenol is one of the key volatile aroma compounds of buckwheat and okra.

Application


  • Anti-Ulcerative Colitis Effects and Active Ingredients in Ethyl Acetate Extract from Decoction of Sargentodoxa cuneata.: This research explores the therapeutic potential of Sargentodoxa cuneata in treating ulcerative colitis, highlighting 2-Methoxy-4-vinylphenol as one of its active ingredients. It offers valuable data for chemists working on new treatments for inflammatory diseases (Yu et al., 2023).

  • Phytochemical characterization, anti-diarrhoeal, analgesic, anti-inflammatory activities and toxicity profile of Ananas comosus (L.) Merr (pineapple) leaf in albino rats.: This comprehensive study assesses the medicinal properties of pineapple leaf, identifying 2-Methoxy-4-vinylphenol among its phytochemicals. It supports further research into its use for pain relief and anti-inflammatory purposes (Ugbogu et al., 2024).

Packaging

Packaged in glass bottles

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ralph Dorfner et al.
Journal of agricultural and food chemistry, 51(19), 5768-5773 (2003-09-04)
The formation of 4-vinylguaiacol, guaiacol, and phenol during coffee roasting was monitored in real-time, using resonance enhanced multiphoton ionization and time-of-flight mass spectrometry. A model is proposed, based on two connected reaction channels. One channel, termed the "low activation energy"
B Karmakar et al.
Journal of biotechnology, 80(3), 195-202 (2000-08-19)
A new strain Bacillus coagulans BK07 was isolated from decomposed wood-bark, based on its ability to grow on ferulic acid as a sole carbon source. This strain rapidly decarboxylated ferulic acid to 4-vinylguaiacol, which was immediately converted to vanillin and
Identification of buckwheat (Fagopyrum esculentum Moench) aroma compounds with GC?MS.
Janes D, et al.
Food Chemistry, 112(1), 120-124 (2009)
Stefan Coghe et al.
Journal of agricultural and food chemistry, 52(3), 602-608 (2004-02-05)
The release of ferulic acid and the subsequent thermal or enzymatic decarboxylation to 4-vinylguaiacol are inherent to the beer production process. Phenolic, medicinal, or clove-like flavors originating from 4-vinylguaiacol frequently occur in beer made with wheat or wheat malt. To
Z Huang et al.
Journal of bacteriology, 176(19), 5912-5918 (1994-10-01)
A ferulic acid decarboxylase enzyme which catalyzes the decarboxylation of ferulic acid to 4-hydroxy-3-methoxystyrene was purified from Pseudomonas fluorescens UI 670. The enzyme requires no cofactors and contains no prosthetic groups. Gel filtration estimated an apparent molecular mass of 40.4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service