Skip to Content
Merck
All Photos(1)

Key Documents

W247804

Sigma-Aldrich

Farnesol

mixture of isomers, ≥95%, stabilized, FG

Synonym(s):

3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
222.37
FEMA Number:
2478
Beilstein:
1763926
EC Number:
Council of Europe no.:
78
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.029
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

contains

α-tocopherol (synthetic stabilizer)

may contain

≤50 ppm hexane

refractive index

n20/D 1.489 (lit.)

bp

149 °C/4 mmHg (lit.)

density

0.886 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fresh; floral; sweet

SMILES string

C/C(CC/C=C(C)/CCC=C(C)C)=C\CO

InChI

1S/C15H26O/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11,16H,5-6,8,10,12H2,1-4H3/b14-9+,15-11+

InChI key

CRDAMVZIKSXKFV-YFVJMOTDSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Farnesol repurposing for prevention and treatment of Acinetobacter baumannii biofilms.: This article explores the efficacy of repurposing farnesol for preventing and treating biofilms caused by Acinetobacter baumannii, highlighting its potential as an antibacterial agent (Tan et al., 2024).

  • Methyl-Jasmonate Functions as a Molecular Switch Promoting Cross-Talk between Pathways for the Biosynthesis of Isoprenoid Backbones Used to Modify Proteins in Plants.: This study investigates the role of methyl-jasmonate in regulating the biosynthesis of isoprenoid backbones, which are crucial for post-translational modification of proteins in plants and may involve farnesol as an intermediate or by-product (Chevalier et al., 2024).

  • Repurposing Farnesol for Combating Drug-Resistant and Persistent Single and Polymicrobial Biofilms.: This article discusses the potential of repurposing farnesol to combat drug-resistant and persistent biofilms, demonstrating its utility in enhancing antibiotic efficacy and addressing antimicrobial resistance (Tan et al., 2024).

Biochem/physiol Actions

Taste at 10 ppm

Other Notes

Natural occurrence: Apricot, orange peel and oil, grapefruit juice, cloves, ginger thyme beef, whiskey basil, papaya, anise seed and balsam of Peru.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

311.0 °F - closed cup

Flash Point(C)

155 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E Fayard et al.
Current opinion in lipidology, 12(2), 113-120 (2001-03-27)
Cholesterol and bile acid metabolism is tightly controlled by nuclear receptors. The liver X receptor, an oxysterol-activated nuclear receptor, limits cholesterol accumulation in the body both by stimulating reverse cholesterol transport and by inhibiting intestinal cholesterol absorption. The liver X
Phillip A Lichtor et al.
Nature chemistry, 4(12), 990-995 (2012-11-24)
Selectivity in the catalytic functionalization of complex molecules is a major challenge in chemical synthesis. The problem is magnified when there are several possible stereochemical outcomes and when similar functional groups occur repeatedly within the same molecule. Selective polyene oxidation
Quorum sensing in dimorphic fungi: farnesol and beyond.
Kenneth W Nickerson et al.
Applied and environmental microbiology, 72(6), 3805-3813 (2006-06-06)
Rossana de Aguiar Cordeiro et al.
Veterinary microbiology, 159(3-4), 375-380 (2012-05-15)
Farnesol is a sesquiterpene alcohol that modulates cell-to-cell communication in Candida albicans. In recent years, several studies have shown that this molecule presents inhibitory effects against non-albicans Candida species, Paracoccidioides brasiliensis and bacteria. The present study aimed at determining the
Efrat Mashiach-Farkash et al.
Oncotarget, 3(6), 629-639 (2012-07-11)
Neurofibromin regulates cell motility via three distinct GTPase pathways acting through two different domains, the Ras GTPase-activating protein-related domain (GRD) and the pre-GRD domain. First, the GRD domain inhibits Ras-dependent changes in cell motility through the mitogen activated protein cascade.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service