Skip to Content
Merck
All Photos(2)

Documents

W243615

Sigma-Aldrich

4-Ethylguaiacol

natural, FG

Synonym(s):

4-Ethyl-2-methoxyphenol, NSC 82313

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H3-2-(OCH3)OH
CAS Number:
Molecular Weight:
152.19
FEMA Number:
2436
EC Number:
Council of Europe no.:
176
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.008
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥97.5%

refractive index

n20/D 1.528 (lit.)

bp

234-236 °C (lit.)

mp

15 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

bacon; clove; leather; spicy; smoky

SMILES string

CCc1ccc(O)c(OC)c1

InChI

1S/C9H12O2/c1-3-7-4-5-8(10)9(6-7)11-2/h4-6,10H,3H2,1-2H3

InChI key

CHWNEIVBYREQRF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

226.4 °F

Flash Point(C)

108 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ileana Vigentini et al.
FEMS yeast research, 8(7), 1087-1096 (2008-06-21)
Contamination of wine by Dekkera/Brettanomyces bruxellensis is mostly due to the production of off-flavours identified as vinyl- and especially ethyl-phenols, but these yeasts can also produce several other spoiling metabolites, such as acetic acid and biogenic amines. Little information is
M Toyoda et al.
Pharmacology, 47(5), 300-308 (1993-11-01)
Wood creosote, a mixture of phenolic compounds, suppresses in vitro contractions of rat intestine. To identify a compound in wood creosote able to inhibit intestinal motility, we screened its constituent phenolic compounds and found 4-ethylguaiacol (4-EG) as an active compound.
C van der Sluis et al.
Journal of biotechnology, 88(2), 129-139 (2001-06-14)
Immobilization of salt-tolerant yeasts considerably decreases the total time required for the flavour development in soy-sauce processes. For immobilization of cells, alginate gel is mostly used as support material. However, alginate is not very suitable for use in soy-sauce processes
Amalia Z Berna et al.
Journal of agricultural and food chemistry, 56(9), 3238-3244 (2008-04-17)
Taints caused by Brettanomyces sp. spoilage are of concern to winemakers and consumers. Typically the taints are described as "barnyard", "sweaty saddle", and "Band-aid" when present in red wine at concentrations of several hundred micrograms per liter or more. The
Iavor Tchobanov et al.
FEMS microbiology letters, 284(2), 213-217 (2008-06-26)
Brettanomyces is the major microbial cause for wine spoilage worldwide and causes significant economic losses. The reasons are the production of ethylphenols that lead to an unpleasant taint described as 'phenolic odour'. Despite its economic importance, Brettanomyces has remained poorly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service