Skip to Content
Merck
All Photos(1)

Documents

W241811

Sigma-Aldrich

Ethyl acrylate

≥99.5%, stabilized

Synonym(s):

Acrylic acid ethyl ester, Ethyl 2-propenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOOC2H5
CAS Number:
Molecular Weight:
100.12
FEMA Number:
2418
Beilstein:
773866
Council of Europe no.:
245
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.037
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Kosher

reg. compliance

FDA 21 CFR 117

vapor density

3.5 (vs air)

vapor pressure

31 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

721 °F

contains

10-30 ppm MEHQ as stabilizer (synthetic)

expl. lim.

12.1 %

refractive index

n20/D 1.406 (lit.)

bp

99 °C (lit.)

mp

−71 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

plastic

SMILES string

CCOC(=O)C=C

InChI

1S/C5H8O2/c1-3-5(6)7-4-2/h3H,1,4H2,2H3

InChI key

JIGUQPWFLRLWPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl acrylate can be used as a flavoring agent in the food industry. It is one of the main volaitle components identified in organic passion fruit pulp.

Application


  • Refractive index adjustable intraocular lens design to achieve diopter control for improving the treatment of ametropia after cataract surgery.: This study involves the use of ethyl acrylate in developing intraocular lenses with adjustable refractive indices, which aims to improve visual outcomes in patients undergoing cataract surgery by providing customizable diopter control (Hong et al., 2024).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of the cultivation system in the aroma of the volatile compounds and total antioxidant activity of passion fruit
Janzantti NS, et al.
LWT--Food Science and Technology, 46(2), 511-518 (2012)
Burdock, GA
Encyclopedia of Food and Color Additives, 1, 945-946 (1997)
M Monier et al.
Journal of hazardous materials, 176(1-3), 348-355 (2009-12-08)
The graft copolymerization of ethyl acrylate (EA) onto natural wool fibers initiated by potassium persulphate and Mohr's salt redox initiator system in limited aqueous medium was carried out in heterogeneous media. Ester groups of the grafted copolymers were partially converted
M Shahjahan Kabir et al.
The Journal of organic chemistry, 77(1), 300-310 (2011-11-15)
The stereospecific synthesis of aryloxy and amino substituted E- and Z-ethyl-3-acrylates is of interest because of their potential in the polymer industry and in medicinal chemistry. During work on a copper-catalyzed cross-coupling reaction of ethyl (E)- and (Z)-3-iodoacrylates with phenols
Anna E V Hagström et al.
Biotechnology and bioengineering, 102(3), 693-699 (2008-09-30)
An OH-functional polyester has been acrylated via transesterification of ethyl acrylate, catalyzed by Candida antarctica lipase B (CalB) in two different preparations: Novozym 435 and immobilized on Accurel MP1000. The batch process resulted in incomplete acrylation as well as severe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service