Skip to Content
Merck
All Photos(3)

Key Documents

W240015

Sigma-Aldrich

γ-Dodecalactone

greener alternative

natural, ≥98%, FG

Synonym(s):

(±)-4-Dodecanolide, (±)-γ-Octyl-γ-butyrolactone, (±)-Dihydro-5-octyl-2(3H)-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
FEMA Number:
2400
Beilstein:
126680
EC Number:
Council of Europe no.:
2240
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.019
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
biological source:
Cocos nucifera
food allergen:
tree nuts

biological source

Cocos nucifera

Quality Level

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

refractive index

n20/D 1.452 (lit.)

bp

130-132 °C/1.5 mmHg (lit.)

mp

17-18 °C (lit.)

density

0.936 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

tree nuts

greener alternative category

Organoleptic

creamy; dairy; fatty; fruity; waxy; peach

SMILES string

CCCCCCCCC1CCC(=O)O1

InChI

1S/C12H22O2/c1-2-3-4-5-6-7-8-11-9-10-12(13)14-11/h11H,2-10H2,1H3

InChI key

WGPCZPLRVAWXPW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased product, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Other Notes

Natural occurrence: Blue cheese, heated butter, pork fat, rum, sherry and cheeses.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service