Skip to Content
Merck
All Photos(1)

Key Documents

W234109

Sigma-Aldrich

Cuminaldehyde

≥98%, FCC, FG

Synonym(s):

4-Isopropylbenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H4CHO
CAS Number:
Molecular Weight:
148.20
FEMA Number:
2341
Beilstein:
636547
EC Number:
Council of Europe no.:
111
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.022
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

300
400

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

composition

contains IFRA restricted Cuminaldehyde

refractive index

n20/D 1.529 (lit.)

bp

235-236 °C (lit.)

solubility

ethanol: 1mL/4 mL, clear, colorless (70%)

density

0.981 g/mL at 20 °C
0.977 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

cuminaldehyde

Organoleptic

spicy; vegetable; warm

SMILES string

[H]C(=O)c1ccc(cc1)C(C)C

InChI

1S/C10H12O/c1-8(2)10-5-3-9(7-11)4-6-10/h3-8H,1-2H3

InChI key

WTWBUQJHJGUZCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

199.4 °F

Flash Point(C)

93 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Désage et al.
Journal of chromatography. B, Biomedical applications, 678(2), 205-210 (1996-04-12)
The flavours contained in a mammalian mother's milk can exert a marked influence on her offspring's proximate suckling behaviour and later preferences. The aim of this study was to establish a reliable analytical procedure to characterise the mammary transfer of
Hoi-Seon Lee
Journal of agricultural and food chemistry, 53(7), 2446-2450 (2005-03-31)
The inhibitory activity of Cuminum cyminum seed-isolated component was evaluated against lens aldose reductase and alpha-glucosidase isolated from Sprague-Dawley male rats and compared to that of 11 commercially available components derived from C. cyminum seed oil, as well as quercitrin
Teruhiko Nitoda et al.
Phytotherapy research : PTR, 22(6), 809-813 (2008-04-17)
Cuminaldehyde (4-isopropylbenzaldehyde) suppressed melanin formation in cultured murine B16-F10 melanoma cells in a dose-dependent decrease up to 0.25 mm without affecting cell growth. Approximately 30% suppression in melanin production resulted when the cells were cultured with 0.25 mm of cuminaldehyde.
T P Krishnakantha et al.
Indian journal of biochemistry & biophysics, 30(2), 133-134 (1993-04-01)
Effect of spice principles on scavenging of superoxide anion has been investigated. The superoxide anions, as measured by nitrobluetetrazolium (NBT) reduction in xanthine-xanthine oxidase system, were inhibited by superoxide dismutase, spice principles eugenol (cloves) and cuminaldehyde (cumin), antioxidants, butylated hydroxy
Qinqin Chen et al.
Analytica chimica acta, 689(1), 149-154 (2011-02-23)
High-speed counter-current chromatography (HSCCC) technique in semi-preparative scale was successfully used in isolation and purification of cuminaldehyde and p-menta-1,4-dien-7-al from the essential oil of Cuminum cyminum L. by using a two-phase solvent system composed of n-hexane-methanol-water (5:4:1, v/v/v). The targeted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service