Skip to Content
Merck
All Photos(1)

Documents

W233706

Sigma-Aldrich

p-Cresol

≥99%, FG

Synonym(s):

4-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
FEMA Number:
2337
Beilstein:
1305151
EC Number:
Council of Europe no.:
619
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.028
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515
FDA

vapor density

3.72 (vs air)

vapor pressure

1 mmHg ( 20 °C)

Assay

≥99%

form

solid or liquid

autoignition temp.

1038 °F

expl. lim.

1 %
1.1 %, 150 °F

bp

202 °C (lit.)

mp

32-34 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

phenolic

SMILES string

Cc1ccc(O)cc1

InChI

1S/C7H8O/c1-6-2-4-7(8)5-3-6/h2-5,8H,1H3

InChI key

IWDCLRJOBJJRNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Cresol is a simple monophenolic aroma compound that occurs naturally in buckwheat honey. It is one of the main volatile compounds responsible for the sheepy-muttony aroma in ovine fats. p-Cresol is also reported to be responsible for the undesirable cowy/barny flavor in nonfat dry milk (NDM).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Food phenolics, pros and cons: a review
Umar Lule S & Xia W
Food Reviews International, 21(4), 367-388 (2005)
Volatile alkylphenols and thiophenol in species?related characterizing flavors of red meats
a J & Lindsay RC
Journal of Food Science, 56(5), 1197-1202 (1991)
Yonca Karagül-Yüceer et al.
Journal of agricultural and food chemistry, 50(2), 305-312 (2002-01-10)
Nonfat dry milk (NDM) is widely used both as an ingredient in other preparations and for direct consumption. Flavor quality of NDM is a critical parameter because it can directly impact final product quality. Flavors can be formed in NDM
Simple phenol and phenolic compounds in food flavor
Maga JA & Katz I.
Critical Reviews in Food Science and Nutrition, 10(4), 323-372 (1978)
Francois Brial et al.
Cell reports, 30(7), 2306-2320 (2020-02-23)
Exposure to natural metabolites contributes to the risk of cardiometabolic diseases (CMDs). Through metabolome profiling, we identify the inverse correlation between serum concentrations of 4-cresol and type 2 diabetes. The chronic administration of non-toxic doses of 4-cresol in complementary preclinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service