Skip to Content
Merck
All Photos(3)

Key Documents

W222011

Sigma-Aldrich

Isobutyraldehyde

greener alternative

natural, 96%, FG

Synonym(s):

2-Methylpropanal, 2-Methylpropionaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCHO
CAS Number:
Molecular Weight:
72.11
FEMA Number:
2220
Beilstein:
605330
EC Number:
Council of Europe no.:
92c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.004
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

300
400

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

vapor density

2.5 (vs air)

vapor pressure

66 mmHg ( 4.4 °C)

Assay

96%

autoignition temp.

384 °F

expl. lim.

10 %, 25 °F
2 %, 32 °F

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

refractive index

n20/D 1.374 (lit.)

bp

63 °C (lit.)

mp

−65 °C (lit.)

solubility

60 g/L at 25 °C

density

0.79 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

cocoa; pungent; brown

SMILES string

[H]C(=O)C(C)C

InChI

1S/C4H8O/c1-4(2)3-5/h3-4H,1-2H3

InChI key

AMIMRNSIRUDHCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Efficient total synthesis of pulchellalactam, a CD45 protein tyrosine phosphatase inhibitor.: Discusses a synthetic route involving isobutyraldehyde for developing inhibitors that have potential applications in biochemistry and medical research (Li WR et al., 2002).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-11.2 °F - closed cup

Flash Point(C)

-24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bettina M Pause et al.
Psychophysiology, 40(2), 209-225 (2003-06-25)
The aim of the present study was to investigate the similarities and differences in the olfactory and visual processing of emotional stimuli in healthy subjects and in patients with major depressive disorder (MDD). Twenty-five inpatients were investigated after admission to
Hyunwoo Kim et al.
Organic letters, 11(1), 157-160 (2008-12-10)
Addition of isobutyraldehyde to 1,2-bis(2-hydroxyphenyl)-1,2-diaminoethane (mother diamine) cleanly gives a stable, fused imidazolidine-dihydro-1,3-oxazine ring complex. However, vigorous heating of the fused ring complex gives the diaza-Cope rearrangement product with excellent yield and stereoselectivity. A variety of alkyl aldehydes have been
Shota Atsumi et al.
Nature biotechnology, 27(12), 1177-1180 (2009-11-17)
Global climate change has stimulated efforts to reduce CO(2) emissions. One approach to addressing this problem is to recycle CO(2) directly into fuels or chemicals using photosynthesis. Here we genetically engineered Synechococcus elongatus PCC7942 to produce isobutyraldehyde and isobutanol directly
Rudy Coquet et al.
Physical chemistry chemical physics : PCCP, 9(45), 6040-6046 (2007-11-16)
The formation and catalytic behavior of active structures in a SiO(2)-supported unsaturated Ru complex catalyst for alkene epoxidation were studied by means of hybrid density functional theory (DFT) calculations. An energy-gaining route for the catalyst activation was found to allow
Hamid Reza Khavasi et al.
Inorganic chemistry, 48(13), 5593-5595 (2009-05-28)
Dioxygen epoxidation of cyclic alkenes into their corresponding epoxides was successfully achieved in good yield by using a novel binuclear manganese carboxamide complex as the catalyst and isobutyraldehyde as the cosubstrate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service