SYNV210250G
Synthware™ Dewar type vacuum trap
250 mL, stopcock: glass
Synonym(s):
Synthware V210250G
All Photos(1)
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Synthware V210250G
capacity
250 mL
Related Categories
General description
Dry ice, Dewar style trap condenser designed for large volume solvent removal. Choice either glass stopcock for better vacuum or PTFE stopcock for grease-free operation. With 10 mm hose connections.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)