Key Documents
SYNC184603
Synthware chromatography column with PTFE stopcock
top joint: ST/NS 24/40, I.D. × L 53.0 mm × 305 mm
Synonym(s):
Synthware C184603, glass chromatograpy column
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C184603
I.D. × L
53.0 mm × 305 mm
suitability
suitable for chromatography
General description
General purpose column has a PTFE stopcock and a top outer joint. For flash chromatography. All Synthware columns are heavy wall designed to increase safety.
This product does not include coarse fritted disc.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)