Key Documents
SYNA232419
Synthware anti-splash adapter with fritted disc
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 19/22
Synonym(s):
Synthware A232419, rotary evaporator bump trap
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
bottom joint: ST/NS19/22
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A232419
capacity
100 mL
Related Categories
General description
With extra coarse fritted disc sealed above lower inner joint. Frit reduces foam from being carryover.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)