SLW1137/08D
Pyrex® Erlenmeyer flask, graduated with SVL screwcaps
capacity 500 mL
Synonym(s):
Pyrex, SciLabware 1137/08D
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PBT cap
PTFE seal
Pyrex flask
flat bottom
Joint
joint neck SVL 42
sterility
non-sterile
feature
graduated
with cap
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 1137/08D
body O.D.
105 mm
cap size
42
capacity
500 mL
flask H
160 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Ideal for storage of culture specimens and media
- PBT screwcaps with PTFE faced sealing disc
- Erlenmeyer pattern
- Manufactured from Pyrex® borosilicate glass for highest resistance to thermal shock and chemical corrosion
- White enamel graduations and marking spot
- Graduated to full capacity
Other Notes
Browse all Erlenmeyer flasks
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service