Skip to Content
Merck
All Photos(1)

Documents

P8602

Sigma-Aldrich

1-Penten-3-ol

99%

Synonym(s):

Ethyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(OH)CH=CH2
CAS Number:
Molecular Weight:
86.13
Beilstein:
1719833
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.424 (lit.)

bp

114-115 °C (lit.)

density

0.838 g/mL at 20 °C (lit.)
0.839 g/mL at 25 °C (lit.)

SMILES string

CCC(O)C=C

InChI

1S/C5H10O/c1-3-5(6)4-2/h3,5-6H,1,4H2,2H3

InChI key

VHVMXWZXFBOANQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Genovese et al.
Journal of the science of food and agriculture, 98(9), 3376-3383 (2017-12-27)
The interindividual variability observed in saliva characteristics raises the question of its relationship with variability in fat sensory perception, particularly in aroma compounds. In the present study, which aimed to measure aroma release from different individuals, eleven key aroma compounds
A M Killary et al.
Experimental cell research, 154(2), 442-453 (1984-10-01)
A selective system toxic towards mammalian cells expressing the liver-specific isozyme of alcohol dehydrogenase (L-ADH) has been developed. A number of alpha-unsaturated primary and secondary alcohols were assayed for their ability to serve as substrates for rat liver ADH and
J J Garrido et al.
Biochemical genetics, 28(9-10), 513-522 (1990-10-01)
The detoxification of 1-pentene-3-ol (pentenol) and 1-pentene-3-one (pentenone) by Drosophila melanogaster adult flies has been studied in two homozygous lines for the AdhF and AdhS alleles (LRC lines), in their respective lines selected for tolerance to ethanol (LRSe lines) and
A J Fisher et al.
Phytochemistry, 62(2), 159-163 (2002-12-17)
Large releases of 1-penten-3-ol (pentenol) and 1-penten-3-one (pentenone) were recently observed from a variety of leaves subjected to freeze-thaw damage in the presence of oxygen. In order to understand the biochemical origins of these volatiles, soybean leaf extracts were used
Ana Rivas-Cañedo et al.
Meat science, 93(2), 178-186 (2012-10-03)
The effect of dietary supplementation with either vitamin E (300 ppm) or a red wine extract rich in polyphenols (900 ppm) in an omega-3 enriched concentrate on the volatile fraction of lamb meat was assessed. The effect of refrigerated storage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service