Skip to Content
Merck
All Photos(1)

Key Documents

P7629

Sigma-Aldrich

N-Phenylthiourea

≥98%

Synonym(s):

1-Phenyl-2-thiourea, PTU, Phenylthiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCSNH2
CAS Number:
Molecular Weight:
152.22
Beilstein:
907309
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98%

form

powder

mp

145-150 °C (lit.)

SMILES string

NC(=S)Nc1ccccc1

InChI

1S/C7H8N2S/c8-7(10)9-6-4-2-1-3-5-6/h1-5H,(H3,8,9,10)

InChI key

FULZLIGZKMKICU-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

General description

N-Phenylthiourea, also known as phenylthiocarbamide (PTC) is an organosulfur thiourea containing a phenyl ring, that is commonly used as a reactant to synthesis N- phenylformamidinoformamidinothiocarbamide.

Application

N-Phenylthiourea can be used as a reactant to synthesize:
  • Thiazoles via solvent- and catalyst-free condensation with halo ketones[1].
  • Formamidine disulfides via oxidation[2].
  • Luminescent copper(I) cyanide-based coordination polymers[3].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvent-free synthesis of luminescent copper (I) coordination polymers with thiourea derivatives
Grifasi F, et al.
Crystal Growth & Design, 15, 2929-2939 (2015)
Oxidative conversion of thiourea and N-substituted thioureas into formamidine disulfides with acidified chloramine-T: a kinetic and mechanistic approach
Shubha J, et al.
Journal of Sulfur Chemistry, 30, 490-499 (2009)
Microwave assisted solvent-free synthesis of N-phenyl-4-(pyridin-4-yl) thiazoles and their drug-likeness studies
Kukade M, et al.
Journal of the Indian Chemical Society, 99, 100509-100509 (2022)
Panagiotis Symvoulidis et al.
Nature methods, 14(11), 1079-1082 (2017-10-03)
A long-standing objective in neuroscience has been to image distributed neuronal activity in freely behaving animals. Here we introduce NeuBtracker, a tracking microscope for simultaneous imaging of neuronal activity and behavior of freely swimming fluorescent reporter fish. We showcase the
Joseph M Chambers et al.
Cell reports, 33(6), 108370-108370 (2020-11-12)
Cilia are microtubule-based organelles that function in a multitude of physiological contexts to perform chemosensing, mechanosensing, and fluid propulsion. The process of ciliogenesis is highly regulated, and disruptions result in disease states termed ciliopathies. Here, we report that peroxisome proliferator-activated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service