Skip to Content
Merck
All Photos(3)

Key Documents

P6513

Sigma-Aldrich

2-Phenylethylamine hydrochloride

≥98%

Synonym(s):

β-Phenylethylamine hydrochloride, Phenethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
157.64
Beilstein:
3624163
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98%

form

powder

mp

220-222 °C (lit.)

SMILES string

Cl.NCCc1ccccc1

InChI

1S/C8H11N.ClH/c9-7-6-8-4-2-1-3-5-8;/h1-5H,6-7,9H2;1H

InChI key

SKHIBNDAFWIOPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Phenylethylamine hydrochloride is a biogenic aromatic amine.[1][2]

Application

  • Multifunctional organic electrolyte additive for batteries: Research outlined a novel application of 2-Phenylethylamine hydrochloride in developing a multifunctional organic electrolyte additive for aqueous zinc ion batteries, enhancing the performance of polyaniline cathodes (Wang et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calix [5] arene-based heteroditopic receptor for 2-phenylethylamine hydrochloride
Gargiulli C, et al.
The Journal of Organic Chemistry, 74(11), 4350-4353 (2009)
Simultaneous analysis of biogenic amines in canned fish by HPLC.
Yen GC & Hsieh CL
Journal of Food Science, 56(1), 158-160 (1991)
Elena Cichero et al.
Chemical biology & drug design, 81(4), 509-516 (2012-08-14)
Trace amine-associated receptor 1 (TAAR1) is a G protein-coupled receptor that belongs to the family of TAAR receptors and responds to a class of compounds called trace amines, such as β-phenylethylamine (β-PEA) and 3-iodothyronamine (T(1)AM). The receptor is known to
Marion Seitter et al.
International journal of food microbiology, 145(2-3), 483-487 (2011-02-22)
In connection with a study on the DNA microarray based detection of genes involved in safety and technologically relevant properties (Seitter (née Resch) et al., 2011), food-associated coagulase-negative staphylococci (CNS) were investigated phenotypically with regard to their ability to bind
Xin-Long Ni et al.
Organic & biomolecular chemistry, 10(23), 4618-4626 (2012-05-17)
The lower rim functionalized hexahomotrioxacalix[3]arene derivatives cone-3 and cone-5 bearing three benzyl and three N,N-diethyl-2-aminoethoxy groups, respectively, were synthesized from triol 1. Their complexation with 2-(3,4-dihydroxyphenyl)ethylamine (dopamine), 5-hydroxytryptamine (serotonin), and 2-phenylethylamine (phenethylamine), which have biologically important activities, has been studied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service