Skip to Content
Merck
All Photos(1)

Documents

O5284

Sigma-Aldrich

1-Octen-3-ol

98%

Synonym(s):

Pentyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)CH=CH2
CAS Number:
Molecular Weight:
128.21
Beilstein:
1744110
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.437 (lit.)

bp

84-85 °C/25 mmHg (lit.)

density

0.837 g/mL at 20 °C
0.83 g/mL at 25 °C (lit.)

SMILES string

CCCCCC(O)C=C

InChI

1S/C8H16O/c1-3-5-6-7-8(9)4-2/h4,8-9H,2-3,5-7H2,1H3

InChI key

VSMOENVRRABVKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Daigo Terutsuki et al.
Sensors (Basel, Switzerland), 20(20) (2020-10-16)
The advent of 3D-printing technologies has had a significant effect on the development of medical and biological devices. Perfusion chambers are widely used for live-cell imaging in cell biology research; however, air-bubble invasion is a pervasive problem in perfusion systems.
Alicia Pérez-Aparicio et al.
Insects, 10(9) (2019-09-05)
We analysed electroantennogram (EAG) responses of male and female adults of the European grapevine moth Lobesiabotrana (Denis et Schiffermüller) (Lepidoptera: Tortricidae) collected as larvae from grapevine (Vitis vinifera L.) and flax-leaved daphne (Daphne gnidium L.). The host-plant odorants tested were
L E Harrup et al.
Journal of medical entomology, 49(1), 112-121 (2012-02-09)
The host kairomones carbon dioxide (CO2) and 1-octen-3-ol elicit a host seeking response in a wide range of haematophagous Diptera. This study investigates the response of Culicoides biting midges (Diptera: Ceratopogonidae) to these cues using field-based experiments at two sites
Scott A Ritchie et al.
Journal of medical entomology, 50(1), 185-194 (2013-02-23)
Mosquitoes often are collected as part of an arbovirus surveillance program. However, trapping and processing of mosquitoes for arbovirus detection is often costly and difficult in remote areas. Most traps, such as the gold standard Center for Disease control light
Atsuko Araki et al.
The Science of the total environment, 423, 18-26 (2012-03-13)
Microbial volatile organic compounds (MVOCs) are a type of VOCs produced by microorganisms. Exposure to 1-octen-3-ol, one of the known MVOCs, has been reported to reduce nasal patency and increase nasal lavage myeloperoxidase, eosinophil cationic proteins, and lysozymes in both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service