All Photos(1)
About This Item
Linear Formula:
H2C=CHC6H4NO2
CAS Number:
Molecular Weight:
149.15
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Assay
96%
form
liquid
refractive index
n20/D 1.584 (lit.)
mp
−5 °C (lit.)
density
1.07 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
[O-][N+](=O)c1cccc(C=C)c1
InChI
1S/C8H7NO2/c1-2-7-4-3-5-8(6-7)9(10)11/h2-6H,1H2
InChI key
SYZVQXIUVGKCBJ-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
224.6 °F - closed cup
Flash Point(C)
107 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yafei Sun et al.
Nature communications, 11(1), 4600-4600 (2020-09-16)
Supported gold nanoparticles are emerging catalysts for heterogeneous catalytic reactions, including selective hydrogenation. The traditionally used supports such as silica do not favor the heterolytic dissociation of hydrogen on the surface of gold, thus limiting its hydrogenation activity. Here we
S A Bright et al.
Oncology reports, 41(5), 3127-3136 (2019-03-22)
The nitrostyrene scaffold was previously identified as a lead target structure for the development of effective compounds targeting Burkitt's lymphoma. The present study aimed to develop these compounds further in haematological malignancies, including chronic lymphocytic leukaemia (CLL). Cellular viability, flow
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service