MT51350012
Mettler-Toledo technical buffer
pH 11, volume 250 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
51191600
NACRES:
NB.79
Recommended Products
sterility
non-sterile
form
liquid
manufacturer/tradename
Mettler-Toledo 51350012
volume
250 mL
pH
11
Looking for similar products? Visit Product Comparison Guide
General description
METTLER TOLEDO certified pH buffer solutions are traceable to NIST primary standards and come with a quality inspection certificate, which guarantees the stated values and traceability. We offer convenient package sizes from single-use 20 mL pouches to 20 L Cubitainers. Our variety packs of pH 4, 7 and 10 buffers are offered in both 20 mL pouches and convenient 500 mL bottles.
Features and Benefits
- High-quality Mettler-Toledo Rainbow I buffers for accurate sensor calibration and maintenance.
- Small volume packaging (single-use sachets) ensures guaranteed fresh solutions and ease of use.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service