Skip to Content
Merck
All Photos(5)

Key Documents

M3205

Sigma-Aldrich

2-Mercaptobenzimidazole

98%

Synonym(s):

1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2S
CAS Number:
Molecular Weight:
150.20
Beilstein:
119867
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

300-304 °C (lit.)

SMILES string

S=C1Nc2ccccc2N1

InChI

1S/C7H6N2S/c10-7-8-5-3-1-2-4-6(5)9-7/h1-4H,(H2,8,9,10)

InChI key

YHMYGUUIMTVXNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Mercaptobenzimidazole is a good corrosion inhibitor as it reduces the corrosion rate of metals.

Application

2-Mercaptobenzimidazole can be used:
  • To prepare 2-benzimidazolylthioacetophenones by reacting with aromatic ketones, which is a key intermediate for the synthesis of thiazolo[3,2-a]benzimidazoles.
  • To synthesize S-arylated 2-mercapto-benzimidazoles via S-arylation with substituted aryl iodides using CuI and 1,10-phenanthroline.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Skin Sens. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S-arylation of mercaptobenzimidazoles using Cu (I) catalysts-experimental and theoretical observations
Sekar R, et al.
Tetrahedron Letters, 52(26), 3347-3352 (2011)
Edésio F C Alcântara et al.
Journal of colloid and interface science, 311(1), 1-7 (2007-04-10)
The compound 2-mercaptobenzimidazole (MBI) was attached onto a silica gel surface by homogeneous and heterogeneous routes. Both silica modification methodologies resulted in similar products, named SiM(hom) and SiM(het), respectively. These materials were characterized by surface area, infrared, thermogravimetry, and 13C
D M Manohar et al.
Water research, 36(6), 1609-1619 (2002-05-09)
The 2-mercaptobenzimidazole loaded natural clay was prepared for the removal of Hg(II) from aqueous media. Adsorption of the metal ions from aqueous solution as a function of solution concentration, agitation time, pH, temperature, ionic strength, particle size of the adsorbent
Kazue Sakemi et al.
Archives of toxicology, 76(12), 682-691 (2002-11-27)
2-Mercaptobenzimidazole (MBI), a rubber antioxidant, is known to exhibit potent thyroid toxicity in rats, whereas its methylated derivatives are much less toxic. To characterize this methyl-substituent effect on the thyroid toxicity of MBI, comparative toxicokinetic analyses have been conducted in
Corrosion inhibition of zinc in phosphoric acid solution by 2-mercaptobenzimidazole
Wang L, et al.
Corrosion Science, 45(4), 677-683 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service