Skip to Content
Merck
All Photos(2)

Documents

M31104

Sigma-Aldrich

α-Methylbenzylamine

99%

Synonym(s):

(±)-α-Methylbenzylamine, alpha-Methylbenzylamine, (±)-1-Phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
636127
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.526 (lit.)

bp

185 °C/756 mmHg (lit.)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

CC(N)c1ccccc1

InChI

1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3

InChI key

RQEUFEKYXDPUSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Methylbenzylamine is a nitrogen source that is widely used as a representative substrate to study the chemo-enzymatic kinetic resolution of primary amines.

Application

α-Methylbenzylamine can be used as a reactant for the synthesis of dihydro-5H-dibenz[c,e]azepinium salts by reacting with racemic biphenol derivatives.
It is used in the synthesis of o-fluorobenzylamines.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemoenzymatic dynamic kinetic resolution of primary amines.
Paetzold, Jens and Backvall, Jan E
Journal of the American Chemical Society, 127(50), 17620-17621 (2005)
Boyeong Kang et al.
Scientific reports, 7(1), 4786-4786 (2017-07-08)
The energy flow during natural photosynthesis is controlled by maintaining the spatial arrangement of pigments, employing helices as scaffolds. In this study, we have developed porphyrin-peptoid (pigment-helix) conjugates (PPCs) that can modulate the donor-acceptor energy transfer efficiency with exceptional precision
Nora Weber et al.
Microbial cell factories, 16(1), 3-3 (2017-01-05)
Whole-cell biocatalysis based on metabolically active baker's yeast with engineered transamination activity can be used to generate molecules carrying a chiral amine moiety. A prerequisite is though to express efficient ω-transaminases and to reach sufficient intracellular precursor levels. Herein, the
Kinetic resolution of ??methylbenzylamine with o?transaminase screened from soil microorganisms: Application of a biphasic system to overcome product inhibition.
Shin, Jong-Shik and Kim, Byung-Gee
Biotechnology and Bioengineering, 55(2), 348-358 (1997)
Palladium-Catalyzed ortho-Selective C-H Fluorination of Oxalyl Amide-Protected Benzylamines.
Chen, Changpeng et al.
The Journal of Organic Chemistry, 80(2), 942-949 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service