All Photos(1)
About This Item
Linear Formula:
CH3OC6H4COC6H5
CAS Number:
Molecular Weight:
212.24
Beilstein:
1104713
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
bp
354-356 °C (lit.)
mp
60-63 °C (lit.)
SMILES string
COc1ccc(cc1)C(=O)c2ccccc2
InChI
1S/C14H12O2/c1-16-13-9-7-12(8-10-13)14(15)11-5-3-2-4-6-11/h2-10H,1H3
InChI key
SWFHGTMLYIBPPA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hui Chen et al.
Chemistry, an Asian journal, 14(6), 781-788 (2018-12-19)
A series of aggregation-induced emission (AIE) fluorescent gelators (TPE-Cn -Chol) were synthesized by attaching tetraphenylethylene (TPE) to cholesterol through an alkyl chain. The properties of the gel, nano-/microaggregate, and condensed phases were studied carefully. TPE-Cn -Chol molecules form AIE fluorescent
Laura Míguez et al.
Ecotoxicology and environmental safety, 208, 111646-111646 (2021-01-06)
Contamination of aquatic ecosystems linked to anthropogenic activity is currently a major concern; therefore, ecotoxicological studies are needed to assess its effect on organisms. The main objective of this study was to investigate the effects of different pollutants on microalgae
J B Rajesha et al.
Water environment research : a research publication of the Water Environment Federation, 89(5), 396-405 (2016-10-26)
Zinc Oxide (ZnO) nanoparticles prepared by gel combustion synthesis using cassava pearl starch have been characterized by various microscopy and spectroscopy techniques. The particle size averaged around 45 nm, revealed zincite structure, hexagonal, agglomerated morphology and possessed an excellent photocatalytic
Thomas William Bentley et al.
International journal of molecular sciences, 12(8), 4805-4818 (2011-09-29)
Rate constants and activations parameters are reported for solvolyses of p-Z-substituted benzoyl chlorides (1, Z = OMe, Me, H, and Cl) in 97% w/w hexafluoroisopropanol-water (97H). Additional kinetic data are reported for solvolyses in acetic and formic acids. Plots of
Rong Zhang et al.
Mikrochimica acta, 186(8), 536-536 (2019-07-19)
The authors describe TiO2 nanowires (TiO2NWs) for solid-phase microextraction of polycyclic aromatic hydrocarbons (PAHs). Well-oriented TiO2NWs were in-situ grown on a titanium wire by a hydrothermal process. The TiO2NWs arrays are radially aligned on the curved titanium fiber substrate. They
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service