Skip to Content
Merck
All Photos(1)

Key Documents

I10406

Sigma-Aldrich

Iodonitrotetrazolium chloride

95%

Synonym(s):

2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, p-Iodonitrotetrazolium Violet, INT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13ClIN5O2
CAS Number:
Molecular Weight:
505.70
Beilstein:
4093224
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

impurities

<5% methyl alcohol

mp

240 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Cl-].[O-][N+](=O)c1ccc(cc1)-[n+]2nc(nn2-c3ccc(I)cc3)-c4ccccc4

InChI

1S/C19H13IN5O2.ClH/c20-15-6-8-16(9-7-15)23-21-19(14-4-2-1-3-5-14)22-24(23)17-10-12-18(13-11-17)25(26)27;/h1-13H;1H/q+1;/p-1

InChI key

JORABGDXCIBAFL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Electron acceptor for the colorimetric assay of various dehydrogenases
Iodonitrotetrazolium chloride (INT) on reduction gets converted to iodonitrotetrazolium formazan, a water insoluble violet colored dye or indicator. Hence INT can be used for the colorimetric assay of various dehydrogenases.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - STOT SE 2 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of cysteine residues of spinach ferredoxin-NADP+ reductase as assessed by site-directed mutagenesis
Aliverti A, et al.
Biochemistry, 32(25), 6374-6380 (1993)
E G Pearson
Journal of the American Veterinary Medical Association, 198(9), 1651-1654 (1991-05-01)
Of 41 equids referred to a veterinary teaching hospital in the Pacific northwest because of dyspnea and inspiratory noise, 3 ponies were diagnosed as having liver failure, most likely attributable to pyrrolizidine alkaloid toxicosis. Dyspnea appeared to be caused by
The measurement of electron transport system activity in river biofilms
Blenkinsopp SA and Lock MA
Water Research, 24(4), 441-445 (1990)
The role of glycerol in the nutrition of halophilic archaeal communities: a study of respiratory electron transport
Oren A
FEMS Microbiology Ecology, 16(4), 281-290 (1995)
F Mermillod-Blondin et al.
Water research, 39(9), 1687-1698 (2005-05-19)
Biogeochemical processes mediated by microorganisms in river sediments (hyporheic sediments) play a key role in river metabolism. Because biogeochemical reactions in the hyporheic zone are often limited to the top few decimetres of sediments below the water-sediment interface, slow filtration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service