Skip to Content
Merck
All Photos(1)

Key Documents

HAM203361

Hamilton® chromatography syringe

for CTC/LEAP Technologies GC Autosampler, model, 701 FN CTC fixed needle, volume × L 10 μL × 51 mm, needle gauge 23s

Synonym(s):

Hamilton Syringe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03
Pricing and availability is not currently available.

material

Microliter syringe

grade

for chromatography

feature

autoclavable: no
model 701 FN CTC fixed needle

manufacturer/tradename

Hamilton

parameter

20-50 °C temp. range (68-122 °F)
2000 psig pressure

needle I.D. × O.D.

0.25 mm × 0.64 mm

needle gauge

23s

volume × L

10 μL × 51 mm

suitability

suitable for chromatography

Looking for similar products? Visit Product Comparison Guide

General description

S-Line syringes deliver exceptional performance for everyday use in HPLC autosampler injections at a great value. The C-Line syringe minimizes the potential for bent needles with a keyed flange design. X-Type syringes are designed for sensitive LC samples (i.e. proteins, peptides, phospholipids, smaller amines, etc.) with a specially coated needle and inner syringe barrel to minimize sample adsorption.

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service