Skip to Content
Merck
All Photos(1)

Documents

H5126

Sigma-Aldrich

2-(4-Hydroxyphenylazo)benzoic acid

≥90% (HPLC)

Synonym(s):

2-(4′-Hydroxybenzeneazo)benzoic acid, 4′-Hydroxyazobenzene-2-carboxylic acid, HABA, HBABA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4N=NC6H4CO2H
CAS Number:
Molecular Weight:
242.23
Beilstein:
1842696
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90% (HPLC)

mp

204-208 °C (lit.)

solubility

ethanol: 20 mg/mL, clear to hazy, orange to very deep orange

SMILES string

Oc1ccc(cc1)\N=N\c2ccccc2C(O)=O

InChI

1S/C13H10N2O3/c16-10-7-5-9(6-8-10)14-15-12-4-2-1-3-11(12)13(17)18/h1-8,16H,(H,17,18)/b15-14+

InChI key

DWQOTEPNRWVUDA-CCEZHUSRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(4-Hydroxyphenylazo)benzoic acid may be used in the preparation of:
  • 2-((4-(3,4-dicyanophenoxy)phenyl)diazenyl)benzoic acid
  • 2,2′-((((4,5-dicyano-1,2-phenylene)bis(oxy))bis(4,1-phenylene))bis(diazene-2,1-diyl)) dibenzoic acid
  • a multifunctional benzoxazine monomer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

New water soluble phenoxy phenyl diazenyl benzoic acid substituted phthalocyanine derivatives: Synthesis, antioxidant activities, atypical aggregation behavior and electronic properties.
Agirtas MS, et al.
Dyes and Pigments, 99(2), 423-431 (2013)
Multifunctional polybenzoxazine nanocomposites containing photoresponsive azobenzene units, catalytic carboxylic acid groups, and pyrene units capable of dispersing carbon nanotubes.
Mohamed MG, et al.
Royal Society of Chemistry Advances, 5(56), 45201-45212 (2015)
Noritaka Kato et al.
The journal of physical chemistry. B, 109(42), 19604-19612 (2006-07-21)
This study reports a homogeneous and competitive fluorescence quenching immunoassay based on gold nanoparticle/polyelectrolyte (Au(NP)/PE) coated latex particles prepared by the layer-by-layer (LbL) technique. First, the resonant energy transfer from a layer of fluorescent PEs to Au(NP) in LbL assembled
Joan-Antoni Farrera et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(7), 2277-2285 (2007-12-29)
The tautomeric equilibria of 2-(4'-hydroxyphenylazo)benzoic acid (HABA) and 2-(3',5'-dimethyl-4'-hydroxyphenylazo)benzoic acid (3',5'-dimethyl-HABA) have been studied by a combination of spectroscopic and computational methods. For neutral HABA in solvents of different polarity (toluene, chloroform, DMSO, DMF, butanol, and ethanol) the azo tautomer
M A Domin et al.
Rapid communications in mass spectrometry : RCM, 13(4), 222-226 (1999-03-31)
The ability to rapidly identify the taxonomic class of the wide variety of microorganisms involved in human and animal disease is becoming increasingly important, especially with the increasing development of resistance to the antibiotics which form the main defence against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service