H3003
2-Heptanol
98%
Synonym(s):
Methyl pentyl carbinol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3(CH2)4CH(OH)CH3
CAS Number:
Molecular Weight:
116.20
Beilstein:
1719088
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
refractive index
n20/D 1.420 (lit.)
bp
160-162 °C (lit.)
density
0.817 g/mL at 25 °C (lit.)
SMILES string
CCCCCC(C)O
InChI
1S/C7H16O/c1-3-4-5-6-7(2)8/h7-8H,3-6H2,1-2H3
InChI key
CETWDUZRCINIHU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
2-Heptanol is an aliphatic secondary alcohol. It can undergo selective oxidation to form the corresponding ketone in the presence of a novel Pd catalyst supported on amphiphilic carbon nanotubes. The distribution coefficient of 2-heptanol between micelles and water indicates that it is solubilized both at the surface and in the interior of the micelles.
Application
2-Heptanol in combination with dimethyl sulfoxide form a porogenic mixture that may be used produce macroporous poly (acrylamide-co-N,N′-methylenebisacrylamide) monolithics via copolymerization.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Eye Irrit. 2 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
138.2 °F - closed cup
Flash Point(C)
59 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Green alcohol oxidation on palladium catalysts supported on amphiphilic hybrid carbon nanotubes.
Benyounes A, et al.
Catalysis Today, 249, 137-144 (2015)
Solubilization of heptanols and a, ?-alkanediols in aqueous solutions of sodium dodecyl sulfate.
Blokhus AM, et al.
Journal of Colloid and Interface Science, 114(1), 9-15 (1986)
Preparation of porous hydrophilic monoliths: Effect of the polymerization conditions on the porous properties of poly (acrylamide-co-N, N'-methylenebisacrylamide) monolithic rods.
Xie S, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 5(6), 1013-1021 (1997)
David J S Patinha et al.
Polymers, 12(9) (2020-08-28)
In this contribution, thin poly(ionic liquid) (PIL) coatings with a well-defined pore structure built up from interpolyelectrolyte complexation between a PIL and poly(acrylic acid) (PAA) were successfully used for enhanced solid phase microextraction (SPME). The introduction of porosity with tunable
David J S Patinha et al.
Talanta, 198, 193-199 (2019-03-17)
In this work, a facile preparation of SPME fibers with increased surface area is presented. The SPME fibers were prepared by grinding poly(ionic liquids) (PILs) to obtain particles of 1-16 µm and, with the aid of a silicon adhesive, attach these
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service