Skip to Content
Merck
All Photos(1)

Key Documents

H1529

Sigma-Aldrich

8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt

≥96%

Synonym(s):

HPTS, Pyranine, Solvent Green 7, Trisodium 8-hydroxypyrene-1,3,6-trisulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H7Na3O10S3
CAS Number:
Molecular Weight:
524.39
Beilstein:
4107272
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

Handling storage condition: Sightly light sensitive

Quality Level

Assay

≥96%

mp

>300 °C (lit.)

fluorescence

λex 454 nm; λem 511 nm (pH 9.1)
λex 454 nm; λem 520 nm in H2O

SMILES string

[Na+].[Na+].[Na+].Oc1cc(c2ccc3c(cc(c4ccc1c2c34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H10O10S3.3Na/c17-11-5-12(27(18,19)20)8-3-4-10-14(29(24,25)26)6-13(28(21,22)23)9-2-1-7(11)15(8)16(9)10;;;/h1-6,17H,(H,18,19,20)(H,21,22,23)(H,24,25,26);;;/q;3*+1/p-3

InChI key

KXXXUIKPSVVSAW-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt (HPTS) can be used to prepare:
  • Extruded fluorescent plastic indicator film for the detection of gaseous and dissolved CO2.
  • Highly fluorescent waterborne polyurethane (WPU) matrix which is used as a pH sensing indicator.
  • Positive (p-type) semiconducting polymeric gels via free radical cross-linking co-polymerization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

481.3 °F

Flash Point(C)

249.6 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Highly CO2 sensitive extruded fluorescent plastic indicator film based on HPTS.
Mills A and Yusufu D
Analyst, 141(3), 999-1008 (2016)
Simone Graf et al.
Scientific reports, 9(1), 4766-4766 (2019-03-20)
The cytochrome bo3 quinol oxidase from Vitreoscilla (vbo3) catalyses oxidation of ubiquinol and reduction of O2 to H2O. Data from earlier studies suggested that the free energy released in this reaction is used to pump sodium ions instead of protons
Aaron C Schmidt et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(8), 1903697-1903697 (2020-04-25)
Trimethylamine (TMA) is a metabolite overtly present in patients suffering from trimethylaminuria (TMAU), a rare genetic disorder characterized by a strong "fishy" body odor. To date, no approved pharmacological treatment to sequester excess TMA on the skin of patients exists.
Zhiping Zhou et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 25(33), 7558-7566 (2005-08-19)
Auditory information is conveyed into the CNS via the spiral ganglion neurons, cells that possess distinctive electrophysiological properties that vary according to their cochlear innervation. Neurons from the base of the cochlea fire action potentials with shorter latencies and durations
8-Hydroxypyrene-1, 3, 6-trisulfonic acid trisodium salt (HPTS) based high fluorescent, pH stimuli waterborne polyurethane coatings.
Kumar R, et al.
Polymer, 136(3), 157-165 (2018)

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service