Skip to Content
Merck
All Photos(2)

Documents

GF97378677

Cerium

foil, light tested, 25x25mm, thickness 0.25mm, as rolled, 99.9%

Synonym(s):

Cerium, CE000220

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ce
CAS Number:
Molecular Weight:
140.12
MDL number:
UNSPSC Code:
12141601
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9%

form

foil

manufacturer/tradename

Goodfellow 973-786-77

resistivity

73 μΩ-cm, 20°C

bp

3443 °C (lit.)

mp

795 °C (lit.)

density

6.67 g/mL at 25 °C (lit.)

SMILES string

[Ce]

InChI

1S/Ce

InChI key

GWXLDORMOJMVQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Van Noorden et al.
Journal of microscopy, 171(Pt 1), 3-16 (1993-07-01)
Methods based on the use of cerium to detect the activity of oxidases and phosphatases are rapidly expanding. Both classes of enzymes can be demonstrated with cerium at the electron and light microscopical level. The in situ detection of H2O2
Flemming R Cassee et al.
Critical reviews in toxicology, 41(3), 213-229 (2011-01-20)
Advances of nanoscale science have produced nanomaterials with unique physical and chemical properties at commercial levels which are now incorporated into over 1000 products. Nanoscale cerium (di) oxide (CeO(2)) has recently gained a wide range of applications which includes coatings
J W McDonald et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 8(8), 859-865 (1995-10-01)
Rare earth pneumoconiosis is an uncommonly reported disease caused by the inhalation of dust containing lanthanides, also known as rare earth metals, which are common industrial materials. The pathologic manifestations and natural history of this disorder are incompletely understood. We
M A Jakupec et al.
Reviews of physiology, biochemistry and pharmacology, 153, 101-111 (2005-01-28)
Cerium is a member of the lanthanide series or rare earth elements which exert diverse biological effects mainly by their resemblance to calcium. This similarity, which is particularly characteristic for the lighter members of the lanthanide series, enables these elements
J P Garner et al.
Burns : journal of the International Society for Burn Injuries, 31(5), 539-547 (2005-06-16)
The introduction of early excision of the burn eschar has contributed to a reduction in burn-related mortality but is not appropriate in all circumstances. Cerium nitrate has been used since 1976, usually in combination with silver sulphadiazine, to improve outcome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service