Skip to Content
Merck
All Photos(2)

Key Documents

GF90687695

Cerium

wire reel, 50mm, diameter 1.0mm, hard, 99.9%

Synonym(s):

Cerium, CE005110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ce
CAS Number:
Molecular Weight:
140.12
MDL number:
UNSPSC Code:
12141601
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9%

form

wire

manufacturer/tradename

Goodfellow 906-876-95

resistivity

73 μΩ-cm, 20°C

bp

3443 °C (lit.)

mp

795 °C (lit.)

density

6.67 g/mL at 25 °C (lit.)

SMILES string

[Ce]

InChI

1S/Ce

InChI key

GWXLDORMOJMVQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endomyocardial fibrosis--the possible connexion with myocardial levels of magnesium and cerium.
S M Valiathan et al.
International journal of cardiology, 28(1), 1-5 (1990-07-01)
Ivana Celardo et al.
Nanoscale, 3(4), 1411-1420 (2011-03-04)
Nanotechnology promises a revolution in pharmacology to improve or create ex novo therapies. Cerium oxide nanoparticles (nanoceria), well-known as catalysts, possess an astonishing pharmacological potential due to their antioxidant properties, deriving from a fraction of Ce(3+) ions present in CeO(2).
C J Van Noorden et al.
Journal of microscopy, 171(Pt 1), 3-16 (1993-07-01)
Methods based on the use of cerium to detect the activity of oxidases and phosphatases are rapidly expanding. Both classes of enzymes can be demonstrated with cerium at the electron and light microscopical level. The in situ detection of H2O2
Flemming R Cassee et al.
Critical reviews in toxicology, 41(3), 213-229 (2011-01-20)
Advances of nanoscale science have produced nanomaterials with unique physical and chemical properties at commercial levels which are now incorporated into over 1000 products. Nanoscale cerium (di) oxide (CeO(2)) has recently gained a wide range of applications which includes coatings
Ali Bumajdad et al.
Advances in colloid and interface science, 147-148, 56-66 (2008-11-26)
This review concerns recent research on the synthesis of cerium oxide (also known as ceria, CeO(2)) in colloidal dispersions media for obtaining high surface area catalyst materials. Nanoparticles as small as 5 nm and surface area as high as 250

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service