Skip to Content
Merck
All Photos(2)

Key Documents

GF89984457

Zirconium

foil, light tested, 100x100mm, thickness 0.05mm, annealed, 99.2%

Synonym(s):

Zirconium, ZR000220

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.2%

form

foil

manufacturer/tradename

Goodfellow 899-844-57

resistivity

40 μΩ-cm, 20°C

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abnormal trace metals in man: zirconium.
H A Schroeder et al.
Journal of chronic diseases, 19(5), 573-586 (1966-05-01)
Oluwatayo F Ikotun et al.
Future medicinal chemistry, 3(5), 599-621 (2011-04-30)
Positron emission tomography, with its high sensitivity and resolution, is growing rapidly as an imaging technology for the diagnosis of many disease states. The success of this modality is reliant on instrumentation and the development of effective and novel targeted
Daryoush Afzali et al.
Environmental monitoring and assessment, 186(6), 3523-3529 (2014-02-04)
In this study, a ternary Zr(IV) system with 2-(5-bromo-2-pyridylazo)-5-diethylaminophenol (5-Br-PADAP) and fluoride was chosen on the basis of dispersive liquid-liquid microextraction method. Zirconium was extracted into the fine droplets of dichlorobenzene as extracting solvent. These drops dispersed as a cloud
Muhammad Shahid et al.
Reviews of environmental contamination and toxicology, 221, 107-127 (2012-10-24)
Zirconium (Zr) is a transition metal that has both stable and radioactive isotopes.This metal has gained significant attention as a major pollutant of concern, partly because it has been prominent in the debate concerning the growing anthropogenic pressure on the
David B N Lee et al.
ASAIO journal (American Society for Artificial Internal Organs : 1992), 56(6), 550-556 (2011-01-20)
Recent years have witnessed a rapid increase in the use of zirconium (Zr)-containing compounds in artificial internal organs. Examples include dental implants and other restorative practices, total knee and hip replacement, and middle-ear ossicular chain reconstruction. In nephrological practice, Zr-containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service