GF80022990
Tantalum
rod, 50mm, diameter 6.35mm, annealed, 99.9%
Synonym(s):
Tantalum, TA007935, Ta
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
vapor pressure
<0.01 mmHg ( 537.2 °C)
Assay
99.9%
form
rod
autoignition temp.
572 °F
manufacturer/tradename
Goodfellow 800-229-90
resistivity
13.5 μΩ-cm, 20°C
L × diam.
50 mm × 6.35 mm
bp
5425 °C (lit.)
mp
2996 °C (lit.)
density
16.69 g/cm3 (lit.)
SMILES string
[Ta]
InChI
1S/Ta
InChI key
GUVRBAGPIYLISA-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The journal of knee surgery, 20(3), 185-194 (2007-08-02)
Porous tantalum represents an alternative metal for primary and revision total knee arthroplasty (TKA) with several unique properties. Tantalum is a transition metal, which in its bulk form has shown excellent biocompatibility and is safe to use in vivo as
Journal of biomedical materials research. Part B, Applied biomaterials, 89(1), 242-251 (2008-10-08)
Conventional porous-coated joint prostheses used in hip and knee reconstruction have demonstrated good clinical results, however, these implants possess some inherent shortcomings such as low volumetric porosity, suboptimal frictional characteristics, and higher modulus of elasticity relative to that of bone.
Zhongguo xiu fu chong jian wai ke za zhi = Zhongguo xiufu chongjian waike zazhi = Chinese journal of reparative and reconstructive surgery, 26(2), 244-247 (2012-03-13)
To review the progress in the treatment of bone defect by porous tantalum implant. Recent literature was extensively reviewed and summarized, concerning the treatment method of bone defect by porous tantalum implant. By right of their unique properties, porous tantalum
Clinical materials, 16(3), 167-173 (1993-12-09)
A detailed literature search was carried out to define the current knowledge about the biological performance of tantalum. The pure metal appears, to a great degree, to be inert both in vivo and in vitro. Both the pure metal and
American journal of orthopedics (Belle Mead, N.J.), 31(4), 216-217 (2002-05-15)
Conventional materials used for orthopedic reconstruction implants limit the design of new and improved implants. Solid metal is rigid. Porous fixation surfaces have low volumetric porosity available for biologic ingrowth and low frictional characteristics for initial implant stability and have
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service