Skip to Content
Merck
All Photos(2)

Key Documents

GF78567654

Holmium

powder, 10g, max. particle size 500 micron, 99.9%

Synonym(s):

Holmium, HO006010

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ho
CAS Number:
Molecular Weight:
164.93
MDL number:
UNSPSC Code:
12141606
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

foil

manufacturer/tradename

Goodfellow 785-676-54

particle size

500 μm

density

8.8 g/mL at 25 °C (lit.)

SMILES string

[Ho]

InChI

1S/Ho

InChI key

KJZYNXUDTRRSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A B Imhoff et al.
Der Orthopade, 27(8), 518-531 (1998-10-21)
The goal for arthroscopic stabilization of anterior glenohumeral instability is to achieve an outcome equivalent to or better than open procedures. A number of arthroscopic procedures have been advocated to reestablish continuity of the inferior glenohumeral ligament complex (IGHLC) with
Rebecca Tooher et al.
The Journal of urology, 171(5), 1773-1781 (2004-04-13)
We undertook a systematic review to assess the safety and efficacy of holmium laser prostatectomy compared to transurethral resection of the prostate. We searched literature databases through August 2002. Holmium laser studies, including holmium laser resection of the prostate (HoLRP)
J N Kabalin et al.
Journal of clinical laser medicine & surgery, 16(1), 21-27 (1998-09-05)
The authors review the current knowledge regarding the application of the Holmium: YAG laser for prostatectomy. Conventional surgical therapies for benign prostatic hyperplasia (BPH) are effective but associated with relatively high morbidity. Laser prostatectomy, using either Neodymium:YAG or potassium-titanyl-phosphate lasers
Ramsay L Kuo et al.
Urology, 62(1), 59-63 (2003-07-03)
To review the complications associated with 206 holmium laser enucleation of the prostate (HoLEP) procedures. HoLEP is a minimally invasive surgical treatment for benign prostatic hyperplasia. A retrospective review was conducted of HoLEPs performed from April 1, 1999 to October
J N Kabalin et al.
Mayo Clinic proceedings, 73(8), 792-797 (1998-08-14)
As alternatives to transurethral resection of the prostate for treatment of benign prostatic hyperplasia, various types of laser prostatectomy-most notably, neodymium:yttrium-aluminum-garnet (YAG) and potassium titanyl phosphate laser procedures-have proved to be associated with considerably less morbidity (for example, bleeding, need

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service