Skip to Content
Merck
All Photos(2)

Key Documents

GF77293592

Bismuth

foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 99.97%

Synonym(s):

Bismuth, BI000120

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
MDL number:
UNSPSC Code:
12141706
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

99.97%

form

foil

manufacturer/tradename

Goodfellow 772-935-92

resistivity

129 μΩ-cm, 20°C

size × thickness

100 x 100 mm × 0.01 mm

bp

1560 °C (lit.)

mp

271 °C (lit.)

density

9.8 g/mL at 25 °C (lit.)

SMILES string

[Bi]

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier P Gisbert et al.
Helicobacter, 10(3), 157-171 (2005-05-21)
To systematically review the Helicobacter pylori eradication efficacy with ranitidine bismuth citrate (RBC) and two antibiotics, and to conduct a meta-analysis of randomized clinical trials comparing the efficacy of proton pump inhibitor (PPI) vs. RBC with two antibiotics for 1
Montserrat Filella
Journal of environmental monitoring : JEM, 12(1), 90-109 (2010-01-19)
Like all elements of the periodic table, bismuth is ubiquitously distributed throughout the environment as a result of natural processes and human activities. It is present as Bi(III) in environmental, biological and geochemical samples. Although bismuth and its compounds are
Hongyan Li et al.
Current opinion in chemical biology, 16(1-2), 74-83 (2012-02-11)
Bismuth has been used in medicine for over two centuries for the treatment of various diseases, in particular for gastrointestinal disorders, owing to its antimicrobial activity. Recent structural characterization of bismuth drugs provides an insight into assembly and pharmacokinetic pathway
Alexander C Ford et al.
World journal of gastroenterology, 14(48), 7361-7370 (2008-12-26)
To assess the safety of bismuth used in Helicobacter pylori (H pylori) eradication therapy regimens. We conducted a systematic review and meta-analysis. MEDLINE and EMBASE were searched (up to October 2007) to identify randomised controlled trials comparing bismuth with placebo
Jorge A R Salvador et al.
Future medicinal chemistry, 4(11), 1495-1523 (2012-08-04)
In recent years, the chemical potential of bismuth and bismuth compounds has been actively exploited. Bismuth salts are known for their low toxicity, making them potential valuable reagents for large-scale synthesis, which becomes more obvious when dealing with products such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service