Skip to Content
Merck
All Photos(2)

Key Documents

GF63991929

Rhenium

wire reel, 0.1m, diameter 1.0mm, annealed, 99.97%

Synonym(s):

Rhenium, RE005140

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Re
CAS Number:
Molecular Weight:
186.21
MDL number:
UNSPSC Code:
12141737
PubChem Substance ID:
NACRES:
NA.23

Assay

99.97%

form

wire

manufacturer/tradename

Goodfellow 639-919-29

L × diam.

0.1 m × 1.0 mm

bp

5596 °C (lit.)
5627 °C (lit.)

mp

3180 °C (lit.)

density

21.02 g/cm3 (lit.)

SMILES string

[ReH]

InChI

1S/Re

InChI key

WUAPFZMCVAUBPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabio Zobi
Chimia, 64(4), 259-265 (2010-12-09)
Transition metal complexes of rhenium and technetium find wide application in nuclear medicine and the chemistry of these elements is still the focus of intense research efforts. For therapeutic and diagnostic applications, currently much attention is dedicated to the development
Francesco Tisato et al.
Mass spectrometry reviews, 23(5), 309-332 (2004-07-21)
Diagnostic nuclear medicine (NM) is among the imaging procedures (together with X-ray, computerized tomography, magnetic resonance, and echography) the clinicians can routinely adopt to image organs or tissues and related disorders. (99m)Tc-based agents are the radiopharmaceuticals of election in diagnostic
John Parnell et al.
Nature communications, 6, 6996-6996 (2015-05-20)
Trace metal data for Proterozoic marine euxinic sediments imply that the expansion of nitrogen-fixing cyanobacteria and diversification of eukaryotes were delayed while the availability of bioessential metals such as molybdenum in the ocean was limited. However, there is increasing recognition
Sahar Hiram-Bab et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(5), 1890-1900 (2015-01-30)
Erythropoietin (EPO) primarily regulates red blood cell formation, and EPO serum levels are increased on hypoxic stress (e.g., anemia and altitude). In addition to anemia, recent discoveries suggest new therapeutic indications for EPO, unrelated to erythropoiesis. We investigated the skeletal
Malcolm Frier
Mini reviews in medicinal chemistry, 4(1), 61-68 (2004-02-03)
The favourable nuclear properties of copper-67 and rhenium-188 for therapeutic application are described, together with methods for the chemical synthesis of a number of derivatives. Survival from invasive bladder cancer has changed little over the past 20 years. The intravesicular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service