GF54792052
Ytterbium
lump, 100g, max. size 25mm, 99.9%
Synonym(s):
Ytterbium, YB006100
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Yb
CAS Number:
Molecular Weight:
173.04
MDL number:
UNSPSC Code:
12141616
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99.9%
form
foil
manufacturer/tradename
Goodfellow 547-920-52
resistivity
28 μΩ-cm, 20°C
max. part. size
25 mm
bp
1196 °C (lit.)
mp
824 °C (lit.)
density
6.54 g/mL at 25 °C (lit.)
SMILES string
[Yb]
InChI
1S/Yb
InChI key
NAWDYIZEMPQZHO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ru Zhou et al.
Journal of nanoscience and nanotechnology, 14(6), 4196-4200 (2014-04-18)
The spectroscopic measurements of orthorhombic CaNb2O6 phosphor doped with Yb3+ ions have been performed. Low temperature spectra indicate that Yb3+ ions may occupy three types of nonequivalent sites inside the CaNb2O6 lattice: a main Yb3+ site dominating the near-infrared emission
Wanyue Shao et al.
Journal of nanoscience and nanotechnology, 14(5), 3887-3892 (2014-04-17)
Water-soluble BaYF5:Yb3+/Er3+ nanoparticles with the surface functionalized by a layer of poly(acrylic acid) (PAA) were synthesized via a facile one-step PAA-assisted hydrothermal method. Bovine serum albumin (BSA) protein was conjugated with BaYF5:Yb3+/Er3+ upconversion nanoparticles via free carboxylic acid groups on
Liming Zhang et al.
Journal of nanoscience and nanotechnology, 14(6), 4710-4713 (2014-04-18)
LiYbF4:Yb, Er nanoparticles have been successfully synthesized by thermal decomposition of multiple trifluoroacetic acid salts. The SEM and TEM results show the size of the LiYF4:Yb, Er nanoparticles is about 100 nm in diagonal line, and the morphology of the
Sethuraman Gayathri et al.
Carbohydrate polymers, 121, 302-308 (2015-02-11)
In this study, chitosan functionalized LaF3:Yb,Er upconverting nanotransducers (UCNTs) with controlled size and shape have been successfully synthesized by a facile one pot precipitation method. The chitosan encapsulated UCNTs show bright upconversion fluorescence upon excitation with 974 nm NIR region.
Li Li et al.
Journal of nanoscience and nanotechnology, 14(6), 4313-4319 (2014-04-18)
Lutetium oxide nanocrystals codoped with Tm3+ and Yb3+ have been successfully synthesized via adjusting the pH values of the precursor solution in a hydrothermal method followed by a subsequent calcination process. The samples were systematically characterized by X-ray diffraction, field-emission
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service