Skip to Content
Merck
All Photos(2)

Key Documents

GF38954076

Titanium

rod, 200mm, diameter 6mm, annealed, 99.6+%

Synonym(s):

Titanium, TI007921

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23

Assay

99.6%

form

rod

autoignition temp.

860 °F

manufacturer/tradename

Goodfellow 389-540-76

resistivity

42.0 μΩ-cm, 20°C

L × diam.

200 mm × 6 mm

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liming Wang et al.
Journal of nanoscience and nanotechnology, 14(6), 4224-4228 (2014-04-18)
Tetragonal Barium titanate (BaTiO3) nanotube arrays have been prepared using the template-assisted hydrothermal method combined with an annealing process. The in-situ chemical conversion of TiO2 nanotube array templates ensured that BaTiO3 maintained the morphology of the nanotube architectures. Moreover, X-ray
Namrata Nataraj et al.
Journal of biomedical nanotechnology, 10(5), 864-870 (2014-04-17)
Staphylococcus aureus is one of the major causes of skin and soft tissue infections. In this study we compared the antimicrobial activity of two different TiO2 nanoformulations against Staphylococcus aureus. We synthesized TiO2 nanoparticles of approximately 80 nm diameter and
Aaron F Cipriano et al.
Journal of biomedical nanotechnology, 10(4), 660-668 (2014-04-17)
The objective of this study was to investigate the adhesion and morphology of bone marrow derived stromal cells (BMSCs) on bulk titanium (Ti) substrates with precisely-patterned surfaces consisting of groove-based gratings with groove widths ranging from 50 micro m down
Wei-Qiang Yu et al.
Journal of nanoscience and nanotechnology, 14(6), 4387-4393 (2014-04-18)
The TiO2 nanotubes by anodization have been extensively studied for medical implant and orthopedic applications because of enhancing bone development. In the present study, a new nano-foveolae structure verified by SEM and AFM was prepared by simulating the nanotubes exfoliation
Shaoming Yang et al.
Journal of nanoscience and nanotechnology, 14(6), 4292-4296 (2014-04-18)
The structural and functional characterizations of titanium nitride (TiN) advanced ceramic microsprings (CMSs), with a coil diameter of several micrometers and synthesized by chemical vapor deposition (CVD) were investigated by microscopy techniques. The CMSs were sufficiently mechanically elastic for extension

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service