GF37916411
Platinum
insulated wire, 2m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Synonym(s):
Platinum, PT005820
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
99.99%
form
wire
manufacturer/tradename
Goodfellow 379-164-11
resistivity
10.6 μΩ-cm, 20°C
L × diam. × thickness
2 m × 0.025 mm × 0.005 mm
bp
3827 °C (lit.)
mp
1772 °C (lit.)
density
21.45 g/cm3 (lit.)
SMILES string
[Pt]
InChI
1S/Pt
InChI key
BASFCYQUMIYNBI-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Platinum-free catalysts as counter electrodes in dye-sensitized solar cells.
ChemSusChem, 5(8), 1343-1357 (2012-08-14)
PloS one, 8(8), e69553-e69553 (2013-08-14)
X-ray repair cross-complementing protein 3 (XRCC3) is an essential gene involved in the double-strand break repair pathway. Published evidence has shown controversial results about the relationship between XRCC3 Thr241Met polymorphism and clinical outcomes of non-small cell lung cancer (NSCLC) patients
Archivos espanoles de urologia, 66(5), 524-535 (2013-06-26)
Cisplatin has been the cornerstone of germ cell testicular tumors therapy since its introduction more tan 30 years ago, and a basic part of the schemes given to multiple ovarian, lung, head and neck, and bladder tumors among others. Some
Accounts of chemical research, 46(8), 1848-1857 (2013-07-03)
An efficient oxygen reduction reaction (ORR) offers the potential for clean energy generation in low-temperature, proton-exchange membrane fuel cells running on hydrogen fuel and air. In the past several years, researchers have developed high-performance electrocatalysts for the ORR to address
Journal of inorganic biochemistry, 114, 106-112 (2012-07-24)
This focused review is devoted to our advances in developing trans platinum complexes as antitumoral complexes, in particular those bearing aliphatic amines. It includes some fundamental aspects of the understanding of non conventional metallodrug mechanism. The perspective is arranged by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service