Skip to Content
Merck
All Photos(2)

Key Documents

GF34764666

Holmium

foil, not light tested, 50x50mm, thickness 0.0125mm, as rolled, 99%

Synonym(s):

Holmium, HO000205

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ho
CAS Number:
Molecular Weight:
164.93
MDL number:
UNSPSC Code:
12141606
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

foil

manufacturer/tradename

Goodfellow 347-646-66

size × thickness

50x50 mm × 0.0125 mm

density

8.8 g/mL at 25 °C (lit.)

SMILES string

[Ho]

InChI

1S/Ho

InChI key

KJZYNXUDTRRSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hemendra N Shah et al.
The Journal of urology, 177(4), 1468-1474 (2007-03-27)
In a prospective manner we evaluated the learning experience of an endourologist inexperienced with holmium laser prostate enucleation and its impact on surgical outcome. We also reviewed the literature to document technical features of holmium laser prostate enucleation at different
Lori B Lerner et al.
The Urologic clinics of North America, 36(4), 485-495 (2009-11-28)
The high-powered holmium laser is an excellent tool for the surgical treatment of benign prostatic hyperplasia. This article discusses the background of holmium use in the prostate and describes the surgical techniques of holmium laser ablation of the prostate and
Hemendra N Shah et al.
BJU international, 100(1), 94-101 (2007-04-11)
To evaluate, in a prospective study, the complications in 280 patients undergoing holmium laser enucleation of the prostate (HoLEP) at our institution, and to review previous reports to determine the overall incidence and types of various complications, and analyse their
A B Imhoff et al.
Der Orthopade, 27(8), 518-531 (1998-10-21)
The goal for arthroscopic stabilization of anterior glenohumeral instability is to achieve an outcome equivalent to or better than open procedures. A number of arthroscopic procedures have been advocated to reestablish continuity of the inferior glenohumeral ligament complex (IGHLC) with
Brian R Matlaga et al.
Current opinion in urology, 17(1), 27-31 (2006-12-05)
The surgical treatment of benign prostatic hyperplasia is a dynamic, evolving field. Holmium laser enucleation of the prostate has been one of the most rigorously analyzed interventions for benign prostatic hyperplasia. In the 12 months since July 2005, a number

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service