GF27305603
Tantalum
wire reel, 2m, diameter 1.0mm, annealed, 99.9%
Synonym(s):
Tantalum, TA005160, Ta
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23
vapor pressure
<0.01 mmHg ( 537.2 °C)
Assay
99.90%
form
wire
autoignition temp.
572 °F
manufacturer/tradename
Goodfellow 273-056-03
resistivity
13.5 μΩ-cm, 20°C
L × diam.
2 m × 1.0 mm
bp
5425 °C (lit.)
mp
2996 °C (lit.)
density
16.69 g/cm3 (lit.)
SMILES string
[Ta]
InChI
1S/Ta
InChI key
GUVRBAGPIYLISA-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jian Lu et al.
Zhongguo xiu fu chong jian wai ke za zhi = Zhongguo xiufu chongjian waike zazhi = Chinese journal of reparative and reconstructive surgery, 26(2), 244-247 (2012-03-13)
To review the progress in the treatment of bone defect by porous tantalum implant. Recent literature was extensively reviewed and summarized, concerning the treatment method of bone defect by porous tantalum implant. By right of their unique properties, porous tantalum
Nilesh Patil et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 89(1), 242-251 (2008-10-08)
Conventional porous-coated joint prostheses used in hip and knee reconstruction have demonstrated good clinical results, however, these implants possess some inherent shortcomings such as low volumetric porosity, suboptimal frictional characteristics, and higher modulus of elasticity relative to that of bone.
J Black
Clinical materials, 16(3), 167-173 (1993-12-09)
A detailed literature search was carried out to define the current knowledge about the biological performance of tantalum. The pure metal appears, to a great degree, to be inert both in vivo and in vitro. Both the pure metal and
Robert Cohen
American journal of orthopedics (Belle Mead, N.J.), 31(4), 216-217 (2002-05-15)
Conventional materials used for orthopedic reconstruction implants limit the design of new and improved implants. Solid metal is rigid. Porous fixation surfaces have low volumetric porosity available for biologic ingrowth and low frictional characteristics for initial implant stability and have
Brett Levine et al.
The journal of knee surgery, 20(3), 185-194 (2007-08-02)
Porous tantalum represents an alternative metal for primary and revision total knee arthroplasty (TKA) with several unique properties. Tantalum is a transition metal, which in its bulk form has shown excellent biocompatibility and is safe to use in vivo as
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service