GF26625067
Holmium
foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99%
Synonym(s):
Holmium, HO000215
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Ho
CAS Number:
Molecular Weight:
164.93
MDL number:
UNSPSC Code:
12141606
PubChem Substance ID:
NACRES:
NA.23
Assay
99%
form
foil
manufacturer/tradename
Goodfellow 266-250-67
size × thickness
50x50 m × 0.05 mm
density
8.8 g/mL at 25 °C (lit.)
SMILES string
[Ho]
InChI
1S/Ho
InChI key
KJZYNXUDTRRSPN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ramsay L Kuo et al.
Urology, 62(1), 59-63 (2003-07-03)
To review the complications associated with 206 holmium laser enucleation of the prostate (HoLEP) procedures. HoLEP is a minimally invasive surgical treatment for benign prostatic hyperplasia. A retrospective review was conducted of HoLEPs performed from April 1, 1999 to October
Rebecca Tooher et al.
The Journal of urology, 171(5), 1773-1781 (2004-04-13)
We undertook a systematic review to assess the safety and efficacy of holmium laser prostatectomy compared to transurethral resection of the prostate. We searched literature databases through August 2002. Holmium laser studies, including holmium laser resection of the prostate (HoLRP)
J N Kabalin et al.
Mayo Clinic proceedings, 73(8), 792-797 (1998-08-14)
As alternatives to transurethral resection of the prostate for treatment of benign prostatic hyperplasia, various types of laser prostatectomy-most notably, neodymium:yttrium-aluminum-garnet (YAG) and potassium titanyl phosphate laser procedures-have proved to be associated with considerably less morbidity (for example, bleeding, need
J N Kabalin et al.
Journal of clinical laser medicine & surgery, 16(1), 21-27 (1998-09-05)
The authors review the current knowledge regarding the application of the Holmium: YAG laser for prostatectomy. Conventional surgical therapies for benign prostatic hyperplasia (BPH) are effective but associated with relatively high morbidity. Laser prostatectomy, using either Neodymium:YAG or potassium-titanyl-phosphate lasers
T A Wollin et al.
Journal of clinical laser medicine & surgery, 16(1), 13-20 (1998-09-05)
To review the physics related to the holmium laser, its laser-tissue interactions, and its application to the treatment of urological diseases. The holmium: YAG laser is a solid-state, pulsed laser that emits light at 2100 nm. It combines the qualities
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service