GF25600777
Silver
foil, 10mm disks, thickness 0.008mm, 99.95+%
Synonym(s):
Silver, AG000218
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23
Assay
99.95%
form
foil
manufacturer/tradename
Goodfellow 256-007-77
resistivity
1.59 μΩ-cm, 20°C
diam. × thickness
10 mm × 0.008 mm
bp
2212 °C (lit.)
mp
960 °C (lit.)
density
10.49 g/cm3 (lit.)
SMILES string
[Ag]
InChI
1S/Ag
InChI key
BQCADISMDOOEFD-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Wen-Wei Zhang et al.
Journal of colloid and interface science, 268(1), 173-180 (2003-11-13)
Self-assembled monolayers (SAMs) of functionalized azobenzene thiols (RAzoCnSH, n=3-6 for R=H, abbreviated as AzoCnSH; and n=4 for R=CH(3)CONH, abbreviated as aaAzoC4SH) on different substrates RAzoCnSz.sbnd;z.sfnc;S (S represents substrates of vacuum-deposited gold (Au), silver foil (Ag), HNO(3) etched silver foil (EAg)
A Rosenblatt et al.
Journal of dental research, 88(2), 116-125 (2009-03-13)
The antimicrobial use of silver compounds pivots on the 100-year-old application of silver nitrate, silver foil, and silver sutures for the prevention and treatment of ocular, surgical, and dental infections. Ag(+) kills pathogenic organisms at concentrations of <50 ppm, and
Zhiping Zhuang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 72(5), 954-958 (2009-02-03)
Surface-enhanced Raman scattering (SERS) of 4,4'-azopyridine (AZPY) on silver foil substrate was measured under 1064nm excitation lines. Density-functional theory (DFT) methods were used to calculate the structure and vibrational spectra of models such as Ag-AZPY, Ag(4)-AZPY and Ag(6)-AZPY complexes with
Mukul Das et al.
Food additives and contaminants, 22(12), 1219-1223 (2005-12-17)
The use of silver foils in various food preparations is a common practice in Middle Eastern and South East Asian countries. The FAO/WHO Joint Expert Committee on Food Additives (JECFA) has included silver in the list of food additives, but
Cheng Yang et al.
Physical chemistry chemical physics : PCCP, 12(43), 14459-14461 (2010-10-01)
This communication describes a new surface-enhanced Raman scattering (SERS) active silver substrate prepared by iodination of the evaporated silver foil. After iodination, the morphology of the silver substrate undergoes a self-evolution process in which it displays accordingly the UV-vis absorption
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service