Skip to Content
Merck
All Photos(2)

Key Documents

GF07832729

Tungsten

foil, 0.2m coil, thickness 0.6mm, coil width 1mm, as rolled, 99.95%

Synonym(s):

Tungsten, W 000335, W

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
MDL number:
UNSPSC Code:
12141747
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 078-327-29

resistivity

4.9 μΩ-cm, 20°C

L × thickness × width

0.2 m × 0.6 mm × 1 mm

bp

5660 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[W]

InChI

1S/W

InChI key

WFKWXMTUELFFGS-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Congshang Wan et al.
Ultramicroscopy, 119, 106-110 (2011-11-15)
Thermionic electron emission from 200 to 500 nm thick coatings of scandium oxide on tungsten foil have been examined in thermionic emission microscopy, spectroscopic photoelectron microcopy, synchrotron radiation and ultraviolet photoelectron spectroscopy (UPS). A clear dependence of the scandium oxide-W
J R Marbach et al.
Physics in medicine and biology, 26(3), 435-443 (1981-05-01)
Curves relating beam energy, scattering foil thickness, central-axis depth-dose, and beam flatness have been generated using data taken on a Siemens 200A betatron. The curve set allows a single combination of tungsten foil thickness and electron beam energy to be
Malcolm H Chisholm et al.
Chemical record (New York, N.Y.), 5(5), 308-320 (2005-10-08)
The reaction of M2(O2CBu(t))4 (M = Mo, W) with a dicarboxylic acid in toluene yields compounds of general formula [M2]-O2C-X-CO2-[M2] ([M2] = M2(O2CBu(t))3; X = conjugated spacer). The M2 units are electronically coupled via interactions between the M2 delta and
A Koutsospyros et al.
Journal of hazardous materials, 136(1), 1-19 (2005-12-14)
Since its discovery, tungsten, a transition element of Group VIb of the Periodic Table of Elements, and its compounds have been considered environmentally benign. Its presence in biological and drinking water samples in Fallon, Nevada, an acute lymphocytic leukemia cluster
José L Domingo
Biological trace element research, 88(2), 97-112 (2002-09-26)
Tungstate is an oxyanion that has biological similarities to vanadate. In recent years, a number of studies have shown the antidiabetic effects of oral tungstate in animal models of diabetes. However, because of the tissue accumulation and potential toxicity derived

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service