G6600
Glycine methyl ester hydrochloride
99%, for peptide synthesis
Synonym(s):
Glycine methyl ester·HCl, Methyl glycinate hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH2CH2COOCH3 · HCl
CAS Number:
Molecular Weight:
125.55
Beilstein:
3593644
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Product Name
Glycine methyl ester hydrochloride, 99%
Quality Level
Assay
99%
form
powder, crystals or chunks
reaction suitability
reaction type: solution phase peptide synthesis
color
white
mp
175 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
Cl.COC(=O)CN
InChI
1S/C3H7NO2.ClH/c1-6-3(5)2-4;/h2,4H2,1H3;1H
InChI key
COQRGFWWJBEXRC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Glycine methyl ester hydrochloride, also known as methyl glycinate hydrochloride, is a derivative of glycine, used in the preparation of amino acids and organic compounds.
Application
Glycine methyl ester hydrochloride is used to synthesize cyclophosphazene compounds with amino acid esters as side groups. Additionally, it can be utilized to synthesize peptides in aqueous medium, which makes it a green method for peptide formation.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A green protocol for peptide bond formation in WEB
M Konwar
Tetrahedron Letters, 57, 2283-2285 (2016)
The investigation of thermosensitive properties of phosphazene derivatives bearing amino acid ester groups
A Uslu
Journal of Molecular Structure, 1136, 90-99 (2017)
Vincenzo Santagada et al.
Journal of combinatorial chemistry, 7(4), 618-621 (2005-07-12)
An easy and convenient microwave-assisted synthesis of N-alkylated glycine methyl esters is described. Parallel and nonparallel combinatorial methods are described and compared. The described reactions are reductive alkylations of several aldehydes and glycine methyl ester in the presence of NaBH3CN.
J M Delfino et al.
International journal of peptide and protein research, 21(4), 440-450 (1983-04-01)
The reactivity of the carboxyl groups in bovine growth hormone was studied by reaction with 1-ethyl-3(3-dimethylaminopropyl) carbodiimide in the presence of an excess of glycinemethylester. Localization in the molecule of the various kinetically distinguishable carboxyl groups was achieved. Highest reactivity
D Levy
Biochimica et biophysica acta, 317(2), 473-481 (1973-08-30)
A procedure has been described for the modification of the amino group of glycine methyl ester, and the amino groups of the hormone, insulin, with the reagent p-nitrophenyl-p-chloromercuribenzoate, effecting the introduction of the chloromercuri group at various defined sites in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service