G3806
Glutaric anhydride
95%
Synonym(s):
Pentanedioic anhydride
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C5H6O3
CAS Number:
Molecular Weight:
114.10
Beilstein:
110051
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
95%
SMILES string
O=C1CCCC(=O)O1
InChI
1S/C5H6O3/c6-4-2-1-3-5(7)8-4/h1-3H2
InChI key
VANNPISTIUFMLH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Esterification mechanism of bagasse modified with glutaric anhydride in 1-allyl-3-methylimidazolium chloride: This study explores the homogenous esterification of bagasse with glutaric anhydride, providing insight into the effects of glutaric anhydride dosage on the process (H Wang et al., 2017).
- Epoxy-anhydride vitrimers from aminoglycidyl resins with high glass transition temperature and efficient stress relaxation: Discusses the use of glutaric anhydride as a hardener in high-performance epoxy-based composites, comparing its effectiveness with other hardeners (M Giebler et al., 2020).
- Effects of glutaric anhydride functionalization on filler-free benzoxazine/epoxy copolymers with shape memory and self-healing properties under near-infrared light: Examines how glutaric anhydride enhances the properties of copolymers, particularly in terms of shape memory and self-healing capabilities (L Amornkitbamrung et al., 2022).
- Surface analysis and thermal behavior of the functionalized cellulose by glutaric anhydride through a solvent-free and catalyst-free process: Investigates the modification of cellulose using glutaric anhydride and analyzes its impact on thermal behavior and surface properties (H Fahim et al., 2023).
- Cyclic anhydrides as powerful tools for bioconjugation and smart delivery: This article reviews the use of glutaric anhydride in bioconjugation, highlighting its potential in creating smart delivery systems for biologically active compounds (MV Spanedda et al., 2021).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
347.0 °F - closed cup
Flash Point(C)
175 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xinli Guo et al.
Molecules (Basel, Switzerland), 25(11) (2020-06-10)
In order to enhance the sensitivity of drug-resistant ovarian cancer cells to cisplatin (DDP), a co-delivery system was designed for simultaneous delivery of curcumin (CUR) and p53 DNA. Firstly, the bifunctional peptide K14 composed of tumor targeting peptide (tLyP-1) and
Dana Luca Motoc et al.
Polymers, 13(4) (2021-02-07)
This contribution focuses on the development of flax and flax/basalt hybrid reinforced composites based on epoxidized linseed oil (ELO) resin, exploiting the feasibility of different ratios of glutaric anhydride (GA) to maleinized linseed oil (MLO) in the hardener system (50:0
W L Shelver et al.
Journal of immunoassay, 21(1), 1-23 (2000-05-11)
Antibody generated from ractopamine-hemiglutarate-KLH was used to develop a ractopamine ELISA. The antibody showed good sensitivity in phosphate buffer, with an IC50 of 4.2 ng/ml (ppb) toward ractopamine and 16.2 ng/ml toward glucuronides of ractopamine conjugated to the phenethanolamine phenol
R L Bowen et al.
Journal of dental research, 79(4), 905-911 (2000-06-01)
Polymerizable cyclodextrin derivatives (PCDs) have been proposed as candidates for use in dental therapeutics (Bowen, 1996; Bowen and Reed, 1997). Here, PCD "libraries" were synthesized by quasi-random reactions of 6 moles of methacrylic anhydride plus 6 moles of cyclic glutaric
Rüdiger Benters et al.
Nucleic acids research, 30(2), E10-E10 (2002-01-15)
The DNA microarray-based analysis of single nucleotide polymorphisms (SNPs) is important for the correlation of genetic variations and individual phenotypes, and for locating disease-causing genes. To facilitate the development of surfaces suitable for immobilization of oligonucleotides, we report here a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service