Skip to Content
Merck
All Photos(1)

Key Documents

G11802

Sigma-Aldrich

2-Guanidinobenzimidazole

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N5
CAS Number:
Molecular Weight:
175.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

mp

242-244 °C (dec.) (lit.)

SMILES string

NC(=N)Nc1nc2ccccc2[nH]1

InChI

1S/C8H9N5/c9-7(10)13-8-11-5-3-1-2-4-6(5)12-8/h1-4H,(H5,9,10,11,12,13)

InChI key

JJWCTKUQWXYIIU-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang Zhao et al.
Communications biology, 4(1), 261-261 (2021-02-28)
Here, we report the identification and characterization of the first proton channels from fungi. The fungal proteins are related to animal voltage-gated Hv channels and are conserved in both higher and lower fungi. Channels from Basidiomycota and Ascomycota appear to
S Friis et al.
The Journal of membrane biology, 184(3), 291-297 (2002-03-14)
In this communication we show that Gd3+ acts as an activator of the apical sodium channel (ENaC) in frog skin epithelia. Application of Gd3+ to the apical solution of frog skin epithelia increased the Na+ absorption measured as the amiloride-inhibitable
M Fronius et al.
The Journal of membrane biology, 195(1), 43-51 (2003-09-23)
In the present study we investigated the effect of extracellular gadolinium on amiloride-sensitive Na(+) current across Xenopus alveolar epithelium by Ussing chamber experiments and studied its direct effect on epithelial Na(+) channels with the patch-clamp method. As observed in various
P Komwatana et al.
The Journal of membrane biology, 162(3), 225-232 (1998-05-16)
We have previously shown that epithelial Na+ channels in mouse mandibular gland duct cells are controlled by cytosolic Na+ and Cl-, acting, respectively, via Go and Gi proteins. Since we found no evidence for control of epithelial Na+ channels by
S D Hillyard et al.
Pflugers Archiv : European journal of physiology, 394(4), 287-293 (1982-10-01)
The abdominal skin of bullfrog larvae (Rana catesbeiana) was placed in an Ussing-type chamber, and its transepithelial electrical parameters were recorded with mucosal solutions of different ionic composition. With "K+-like" cations (K+, NH+4, RB+, Cs+) the power spectra of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service