Documents
ESYNTH020
Sigma-Aldrich
SynLectro Boron-doped Diamond Electrode
size 3 mm × 20 mm × 60 mm
Synonym(s):
Boron-Doped Diamond Electrochemical Electrode
About This Item
UNSPSC Code:
81141502
NACRES:
NB.61
size
3 mm × 20 mm × 60 mm
General description
SynLectro Boron-doped Diamond Electrode is a 3x20x60mm electrode designed for use with the SynLectro Electrolysis Platform for synthetic electrochemistry.
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
Visit the Document Library
Related Content
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.