Skip to Content
Merck
All Photos(1)

Key Documents

CDS002679

Sigma-Aldrich

2-Amino-5-phenylpyridine

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10N2
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

Nc1ccc(cn1)-c2ccccc2

InChI

1S/C11H10N2/c12-11-7-6-10(8-13-11)9-4-2-1-3-5-9/h1-8H,(H2,12,13)

InChI key

OAPVIBHQRYFYSE-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyu Yin et al.
Meat science, 172, 108360-108360 (2020-11-08)
The heterocyclic aromatic amine (HAA) level and quality characteristics of selected Harbin red sausages in the northern Chinese market and the possible differences between traditional sausages and conventional sausages were evaluated in this study. Four varieties of traditional sausages and
Lang Zhang et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 135, 110931-110931 (2019-11-05)
This study evaluated the heterocyclic aromatic amine (HAA) contents and quality characteristics of seven kinds of traditional smoked and roasted poultry products on the northern Chinese market. Harbin smoked chicken had the most abundant total HAAs, followed by Haroulian roasted
Jing Chen et al.
Food & function, 8(11), 3938-3950 (2017-09-22)
The effects of various levels of chili pepper, Sichuan pepper, and black pepper on the amounts of 17 heterocyclic amines (HAs) from seven categories of both free and protein-bound states in roast beef patties were assessed by ultra-performance liquid chromatography-tandem
Shihui Shan et al.
Food chemistry, 337, 127630-127630 (2020-08-14)
The current study developed a cheap and effective method for the simultaneous extraction of 14 heterocyclic aromatic amines (HAAs) in food matrix. Core-shell Fe3O4@PDA nanoparticles were constructed and acted as the magnetic solid-phase extraction adsorbent to separate and purify HAAs
Jing Chen et al.
Journal of agricultural and food chemistry, 65(22), 4493-4499 (2017-05-18)
The effect of different roasting temperatures on the amounts of 17 heterocyclic amines (HAs) from seven categories of both free and protein-bound states in roast beef patties was assessed using an ultraperformance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS) method. There were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service